Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451393_m1
          See other TTC9C GE Assays ›
          Gene Symbol:
          Ttc9c
          Gene Name:
          tetratricopeptide repeat domain 9C
          Gene Aliases:
          RGD1359253
          Chromosome Location:
          Chr.1: 225003859 - 225014869 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451393_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttc9c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007693.1 NP_001007694.1 2 - 3 492 107
          XM_006230984.2 XP_006231046.1 2 - 3 721 107
          XM_006230985.2 XP_006231047.1 3 - 4 637 107
          XM_006230986.2 XP_006231048.1 2 - 3 541 107
          XM_006230987.2 XP_006231049.1 2 - 3 476 107
          XM_006230988.2 XP_006231050.1 3 - 4 508 107
          GenBank mRNA BC062803.1 - 2 - 3 492 6890943 107
          FQ211550.1 - 2 - 3 501 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007221.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ttc9c

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          FK506 binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104114 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104114 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104114 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P552977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P552986 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44240 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76794 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728554 - -
          Affymetrix Rat Gene 1.0 ST Array 10728552 - -
          Affymetrix Rat Gene 1.0 ST Array 10728553 - -
          Affymetrix Rat Gene 1.0 ST Array 10728550 - -
          Affymetrix Rat Gene 1.0 ST Array 10728551 - -
          Affymetrix Rat Gene 1.1 ST Array 10728549 - -
          Rat Genome 230 2.0 Array 1376613_PM_at - -
          Rat Genome 230 2.0 Array 1376613_at - -
          Rat Genome 230 2.0 Array 1383730_at - -
          Rat Genome 230 2.0 Array 1378964_PM_at - -
          Rat Genome 230 2.0 Array 1395375_PM_at - -
          Rat Genome 230 2.0 Array 1378964_at - -
          Rat Genome 230 2.0 Array 1395375_at - -
          Rat Genome 230 2.0 Array 1383730_PM_at - -
          Affymetrix HT Rat Focus Array 1376613_PM_at - -
          Affymetrix HT Rat Focus Array 1376613_at - -
          Affymetrix HT Rat Focus Array 1395375_PM_at - -
          Affymetrix HT Rat Focus Array 1395375_at - -
          Affymetrix Rat Clariom S Assay PSR0100058304.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058298.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058302.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058306.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058307.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058308.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058300.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058301.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058296.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058297.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058303.rn.1 TC0100007221.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058299.rn.1 TC0100007221.rn.1 RTA TC
          Rat Expression Set 230 1376613_at - -
          Rat Expression Set 230 1376613_PM_at - -
          Rat Expression Set 230 1378964_at - -
          Rat Expression Set 230 1395375_at - -
          Rat Expression Set 230 1383730_PM_at - -
          Rat Expression Set 230 1383730_at - -
          Rat Expression Set 230 1395375_PM_at - -
          Rat Expression Set 230 1378964_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728549 - -
          Rat Genome U34 Set rc_AI045484_at - -
          Rat Genome U34 Set rc_AI172018_at - -
          Rat Genome U34 Set rc_AI112973_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22108718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline