Search
Search
| Gene Symbol: Doxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199291.1 | NP_954985.1 | 2 - 3 | 1842 | 90 | |
| XM_006236462.1 | XP_006236524.1 | 3 - 4 | 1868 | 90 | ||
| GenBank mRNA | BC098900.1 | - | 3 - 4 | 1874 | 7443671 | 90 |
Set Membership: |
|