Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451849_g1
          See other AOC1 GE Assays ›
          Gene Symbol:
          Aoc1
          Gene Name:
          amine oxidase, copper containing 1
          Gene Aliases:
          Abp, Abp1
          Chromosome Location:
          Chr.4: 78496043 - 78515585 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451849_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aoc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022935.2 NP_075224.2 3 - 4 2032 65
          XM_006236471.2 XP_006236533.1 3 - 4 2054 65
          XM_006236472.2 XP_006236534.1 3 - 4 1875 65
          GenBank mRNA BC100144.1 - 3 - 4 2032 7460184 65
          FQ220353.1 - 1 - 2 159 65
          FQ228794.1 - 2 - 3 666 65
          FQ230036.1 - 1 - 2 159 65
          X73911.1 - 3 - 4 1914 65
          X73912.1 - 2 - 3 335 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000869.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aoc1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          biological_process
          amine metabolic process
          cellular response to copper ion starvation
          response to drug
          response to antibiotic
          oxidation-reduction process
          cellular response to copper ion
          cellular response to histamine
          cellular response to azide
          cation transmembrane transport
          receptor activity
          cation channel activity
          copper ion binding
          calcium ion binding
          primary amine oxidase activity
          drug binding
          heparin binding
          zinc ion binding
          protein complex binding
          protein homodimerization activity
          quinone binding
          diamine oxidase activity
          histamine oxidase activity
          methylputrescine oxidase activity
          propane-1,3-diamine oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P592385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81731 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P592385 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P592385 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P592385 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P592385 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702142449 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93493 - -
          Agilent Rat cDNA Microarray (G4105A) 702142449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855427 - -
          Affymetrix Rat Gene 1.0 ST Array 10855429 - -
          Affymetrix Rat Gene 1.0 ST Array 10855428 - -
          Affymetrix Rat Gene 1.0 ST Array 10855430 - -
          Affymetrix Rat Gene 1.0 ST Array 10855425 - -
          Affymetrix Rat Gene 1.1 ST Array 10855424 - -
          Rat Genome 230 2.0 Array 1368413_at - -
          Rat Genome 230 2.0 Array 1368413_PM_at - -
          Affymetrix HT Rat Focus Array 1368413_PM_at - -
          Affymetrix HT Rat Focus Array 1368413_at - -
          Affymetrix Rat Clariom S Assay PSR0400006230.rn.1 TC0400000869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006225.rn.1 TC0400000869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006234.rn.1 TC0400000869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006224.rn.1 TC0400000869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006233.rn.1 TC0400000869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006232.rn.1 TC0400000869.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006235.rn.1 TC0400000869.rn.1 RTA TC
          Rat Expression Set 230 1368413_at - -
          Rat Expression Set 230 1368413_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855424 - -
          Rat Genome U34 Set X73911_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21016992 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373859 - -
          RatRef-12 Expression BeadChip ILMN_67191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline