Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451864_m1
          See other RGD1564712 GE Assays ›
          Gene Symbol:
          RGD1564712
          Gene Name:
          RGD1564712
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 79324326 - 79349905 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451864_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1564712
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106931.1 NP_001100401.1 2 - 3 183 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002862.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P161571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161571 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161571 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P368202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83089 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0400020418.rn.1 TC0400002862.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21248043 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354058 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline