Search
Search
| Gene Symbol: S100a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012618.2 | NP_036750.1 | 2 - 3 | 200 | 145 | |
| XM_006232592.2 | XP_006232654.1 | 2 - 3 | 260 | 145 | ||
| GenBank mRNA | FQ215053.1 | - | 2 - 3 | 200 | 145 | |
| FQ220722.1 | - | 2 - 3 | 200 | 145 | ||
| FQ221062.1 | - | 2 - 3 | 200 | 145 | ||
| FQ221355.1 | - | 2 - 3 | 200 | 145 | ||
| FQ221406.1 | - | 2 - 3 | 201 | 145 | ||
| FQ221428.1 | - | 2 - 3 | 200 | 145 | ||
| FQ221508.1 | - | 2 - 3 | 200 | 145 | ||
| FQ221699.1 | - | 2 - 3 | 203 | 145 | ||
| FQ221706.1 | - | 2 - 3 | 201 | 145 | ||
| FQ221757.1 | - | 2 - 3 | 201 | 145 | ||
| FQ221860.1 | - | 2 - 3 | 201 | 145 | ||
| FQ222027.1 | - | 2 - 3 | 201 | 145 | ||
| FQ222069.1 | - | 2 - 3 | 200 | 145 | ||
| FQ222186.1 | - | 2 - 3 | 200 | 145 | ||
| FQ222219.1 | - | 2 - 3 | 201 | 145 | ||
| FQ222585.1 | - | 2 - 3 | 200 | 145 | ||
| FQ222875.1 | - | 2 - 3 | 200 | 145 | ||
| FQ223009.1 | - | 2 - 3 | 200 | 145 | ||
| FQ223047.1 | - | 2 - 3 | 204 | 145 | ||
| FQ223108.1 | - | 2 - 3 | 183 | 145 | ||
| FQ223128.1 | - | 2 - 3 | 201 | 145 | ||
| FQ228364.1 | - | 2 - 3 | 202 | 145 | ||
| FQ228506.1 | - | 2 - 3 | 200 | 145 | ||
| FQ228607.1 | - | 2 - 3 | 202 | 145 | ||
| FQ229007.1 | - | 2 - 3 | 199 | 145 | ||
| FQ229874.1 | - | 2 - 3 | 200 | 145 | ||
| J03628.1 | - | 2 - 3 | 201 | 145 | ||
| X64022.1 | - | 1 - 2 | 143 | 145 | ||
Set Membership: |
|