Search
Search
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014254.1 | NP_001014276.1 | 8 - 9 | 1192 | 156 | |
| XM_008761185.1 | XP_008759407.1 | 8 - 9 | 1397 | 156 | ||
| XM_008761186.1 | XP_008759408.1 | 9 - 10 | 1264 | 156 | ||
| XM_008761187.1 | XP_008759409.1 | 8 - 9 | 1394 | 156 | ||
| XM_008761188.1 | XP_008759410.1 | 8 - 9 | 1376 | 156 | ||
| XM_008761189.1 | XP_008759411.1 | 8 - 9 | 1373 | 156 | ||
| XM_008761190.1 | XP_008759412.1 | 8 - 9 | 1373 | 156 | ||
| XM_008761191.1 | XP_008759413.1 | 8 - 9 | 1370 | 156 | ||
| XM_008761192.1 | XP_008759414.1 | 8 - 9 | 1243 | 156 | ||
| XM_008761193.1 | XP_008759415.1 | 5 - 6 | 1044 | 156 | ||
| XM_008761194.1 | XP_008759416.1 | 5 - 6 | 523 | 156 | ||
| GenBank mRNA | BC083788.1 | - | 8 - 9 | 1192 | 7098251 | 156 |
Set Membership: |
|