Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01452066_g1
          See other MSTO1 GE Assays ›
          Gene Symbol:
          Msto1
          Gene Name:
          misato 1, mitochondrial distribution and morphology regulator
          Gene Aliases:
          RGD1306110
          Chromosome Location:
          Chr.2: 188212211 - 188216482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01452066_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Msto1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106443.1 NP_001099913.1 4 - 5 413 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004051.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          mitochondrion organization
          mitochondrion distribution

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152811 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152811 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152811 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1037523 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824363 - -
          Affymetrix Rat Gene 1.0 ST Array 10824365 - -
          Affymetrix Rat Gene 1.0 ST Array 10824372 - -
          Affymetrix Rat Gene 1.0 ST Array 10824367 - -
          Affymetrix Rat Gene 1.0 ST Array 10824368 - -
          Affymetrix Rat Gene 1.0 ST Array 10824361 - -
          Affymetrix Rat Gene 1.0 ST Array 10824370 - -
          Affymetrix Rat Gene 1.0 ST Array 10824364 - -
          Affymetrix Rat Gene 1.0 ST Array 10824371 - -
          Affymetrix Rat Gene 1.0 ST Array 10824369 - -
          Affymetrix Rat Gene 1.0 ST Array 10824360 - -
          Affymetrix Rat Gene 1.0 ST Array 10824359 - -
          Affymetrix Rat Gene 1.0 ST Array 10824358 - -
          Affymetrix Rat Gene 1.0 ST Array 10824366 - -
          Affymetrix Rat Gene 1.0 ST Array 10824362 - -
          Affymetrix Rat Gene 1.1 ST Array 10824357 - -
          Rat Genome 230 2.0 Array 1385819_PM_at - -
          Rat Genome 230 2.0 Array 1398110_at - -
          Rat Genome 230 2.0 Array 1384538_PM_at - -
          Rat Genome 230 2.0 Array 1385819_at - -
          Rat Genome 230 2.0 Array 1384538_at - -
          Rat Genome 230 2.0 Array 1398110_PM_at - -
          Affymetrix HT Rat Focus Array 1384538_PM_at - -
          Affymetrix HT Rat Focus Array 1385819_at - -
          Affymetrix HT Rat Focus Array 1385819_PM_at - -
          Affymetrix HT Rat Focus Array 1384538_at - -
          Affymetrix Rat Clariom S Assay PSR0200027440.rn.1 TC0200004051.rn.1 RTA TC
          Rat Expression Set 230 1384538_PM_at - -
          Rat Expression Set 230 1385819_PM_at - -
          Rat Expression Set 230 1385819_at - -
          Rat Expression Set 230 1384538_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824357 - -
          Rat Genome U34 Set rc_AI012273_f_at - -
          Rat Genome U34 Set rc_AI072702_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21219551 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361759 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline