Search
Search
| Gene Symbol: Rsrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014172.1 | NP_001014194.1 | 3 - 4 | 474 | 93 | |
| XM_008761054.1 | XP_008759276.1 | 3 - 4 | 441 | 93 | ||
| XM_008761055.1 | XP_008759277.1 | 3 - 4 | 464 | 93 | ||
| GenBank mRNA | BC087665.1 | - | 3 - 4 | 474 | 7317835 | 93 |
Set Membership: |
|