Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01452373_m1
          See other TRPV6 GE Assays ›
          Gene Symbol:
          Trpv6
          Gene Name:
          transient receptor potential cation channel, subfamily V, member 6
          Gene Aliases:
          CaT1, Ecac2, Otrpc3
          Chromosome Location:
          Chr.4: 70918627 - 70934291 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01452373_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpv6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053686.1 NP_446138.1 12 - 13 1802 66
          XM_006236375.2 XP_006236437.1 12 - 13 1418 66
          GenBank mRNA AF160798.1 - 12 - 13 1802 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002771.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpv6

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          calcium ion import
          calcium ion transmembrane transport
          calcium activated cation channel activity
          calcium channel activity
          calmodulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P494274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142353 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P494274 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P494274 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P494274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P494274 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700323838 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16902 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37518 - -
          Agilent Rat cDNA Microarray (G4105A) 700323838 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862213 - -
          Affymetrix Rat Gene 1.0 ST Array 10862214 - -
          Affymetrix Rat Gene 1.0 ST Array 10862210 - -
          Affymetrix Rat Gene 1.0 ST Array 10862211 - -
          Affymetrix Rat Gene 1.0 ST Array 10862207 - -
          Affymetrix Rat Gene 1.0 ST Array 10862201 - -
          Affymetrix Rat Gene 1.0 ST Array 10862204 - -
          Affymetrix Rat Gene 1.0 ST Array 10862212 - -
          Affymetrix Rat Gene 1.0 ST Array 10862203 - -
          Affymetrix Rat Gene 1.0 ST Array 10862205 - -
          Affymetrix Rat Gene 1.0 ST Array 10862209 - -
          Affymetrix Rat Gene 1.0 ST Array 10862206 - -
          Affymetrix Rat Gene 1.0 ST Array 10862208 - -
          Affymetrix Rat Gene 1.0 ST Array 10862202 - -
          Affymetrix Rat Gene 1.1 ST Array 10862200 - -
          Rat Genome 230 2.0 Array 1387565_at - -
          Rat Genome 230 2.0 Array 1398521_PM_at - -
          Rat Genome 230 2.0 Array 1398521_at - -
          Rat Genome 230 2.0 Array 1387565_PM_at - -
          Affymetrix HT Rat Focus Array 1387565_PM_at - -
          Affymetrix HT Rat Focus Array 1387565_at - -
          Affymetrix Rat Clariom S Assay PSR0400019756.rn.1 TC0400002771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400019759.rn.1 TC0400002771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400019757.rn.1 TC0400002771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400019754.rn.1 TC0400002771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400019755.rn.1 TC0400002771.rn.1 RTA TC
          Rat Expression Set 230 1387565_at - -
          Rat Expression Set 230 1387565_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862200 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21856854 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373456 - -
          RatRef-12 Expression BeadChip ILMN_52938 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0