Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01452384_m1
          See other TTC26 GE Assays ›
          Gene Symbol:
          Ttc26
          Gene Name:
          tetratricopeptide repeat domain 26
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 66090178 - 66147998 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01452384_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttc26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025045.1 NP_001020216.1 17 - 18 1664 108
          XM_006236330.2 XP_006236392.1 15 - 16 1542 108
          GenBank mRNA BC085935.1 - 17 - 18 1664 7115235 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000660.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ttc26

          Gene Ontology Categories:

          Function(s)

          smoothened signaling pathway
          protein transport
          intraciliary transport
          cilium assembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52801 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129415 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85353 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098314 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098314 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098314 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P837142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76322 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49513 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854599 - -
          Affymetrix Rat Gene 1.0 ST Array 10854593 - -
          Affymetrix Rat Gene 1.0 ST Array 10854597 - -
          Affymetrix Rat Gene 1.0 ST Array 10854590 - -
          Affymetrix Rat Gene 1.0 ST Array 10854600 - -
          Affymetrix Rat Gene 1.0 ST Array 10854596 - -
          Affymetrix Rat Gene 1.0 ST Array 10854592 - -
          Affymetrix Rat Gene 1.0 ST Array 10854595 - -
          Affymetrix Rat Gene 1.0 ST Array 10854598 - -
          Affymetrix Rat Gene 1.0 ST Array 10854591 - -
          Affymetrix Rat Gene 1.0 ST Array 10854594 - -
          Affymetrix Rat Gene 1.1 ST Array 10854589 - -
          Affymetrix Rat Clariom S Assay PSR0400004763.rn.1 TC0400000660.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400004761.rn.1 TC0400000660.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400004762.rn.1 TC0400000660.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10854589 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20699323 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371694 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline