Search
Search
| Gene Symbol: Thop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172075.2 | NP_742072.2 | 3 - 4 | 512 | 70 | |
| XM_006240995.2 | XP_006241057.1 | 3 - 4 | 532 | 70 | ||
| GenBank mRNA | BC081706.1 | - | 3 - 4 | 439 | 7111806 | 70 |
| M61142.1 | - | 3 - 4 | 435 | 70 | ||
Set Membership: |
|