Search
Search
| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108737.1 | NP_001102207.1 | 10 - 11 | 996 | 100 | |
| XM_006240977.2 | XP_006241039.1 | 10 - 11 | 1231 | 100 | ||
| XM_006240978.1 | XP_006241040.1 | 10 - 11 | 1018 | 100 | ||
Set Membership: |
|