Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01452854_m1
          See other ABCC3 GE Assays ›
          Gene Symbol:
          Abcc3
          Gene Name:
          ATP-binding cassette, subfamily C (CFTR/MRP), member 3
          Gene Aliases:
          Mlp2, Mrp3
          Chromosome Location:
          Chr.10: 82047308 - 82116669 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01452854_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080581.1 NP_542148.1 30 - 31 4509 63
          GenBank mRNA AB010467.1 - 30 - 31 4509 63
          AF072816.1 - 30 - 31 4521 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003587.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcc3

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          drug transmembrane transport
          metabolic process
          response to organic substance
          response to organonitrogen compound
          response to organic cyclic compound
          canalicular bile acid transport
          drug transport
          response to estradiol
          response to lipopolysaccharide
          xenobiotic transport
          transmembrane transport
          ATP binding
          xenobiotic-transporting ATPase activity
          drug transmembrane transporter activity
          bile acid-exporting ATPase activity
          ATPase activity, coupled to transmembrane movement of substances

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158080 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127421 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033761 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033761 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P221974 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P572725 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700307508 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100297 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12902 - -
          Agilent Rat cDNA Microarray (G4105A) 700307508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746297 - -
          Affymetrix Rat Gene 1.0 ST Array 10746309 - -
          Affymetrix Rat Gene 1.0 ST Array 10746320 - -
          Affymetrix Rat Gene 1.0 ST Array 10746308 - -
          Affymetrix Rat Gene 1.0 ST Array 10746314 - -
          Affymetrix Rat Gene 1.0 ST Array 10746312 - -
          Affymetrix Rat Gene 1.0 ST Array 10746300 - -
          Affymetrix Rat Gene 1.0 ST Array 10746323 - -
          Affymetrix Rat Gene 1.0 ST Array 10746304 - -
          Affymetrix Rat Gene 1.0 ST Array 10746305 - -
          Affymetrix Rat Gene 1.0 ST Array 10746301 - -
          Affymetrix Rat Gene 1.0 ST Array 10746325 - -
          Affymetrix Rat Gene 1.0 ST Array 10746326 - -
          Affymetrix Rat Gene 1.0 ST Array 10746302 - -
          Affymetrix Rat Gene 1.0 ST Array 10746310 - -
          Affymetrix Rat Gene 1.0 ST Array 10746299 - -
          Affymetrix Rat Gene 1.0 ST Array 10746295 - -
          Affymetrix Rat Gene 1.0 ST Array 10746294 - -
          Affymetrix Rat Gene 1.0 ST Array 10746321 - -
          Affymetrix Rat Gene 1.0 ST Array 10746306 - -
          Affymetrix Rat Gene 1.0 ST Array 10746298 - -
          Affymetrix Rat Gene 1.0 ST Array 10746322 - -
          Affymetrix Rat Gene 1.0 ST Array 10746313 - -
          Affymetrix Rat Gene 1.0 ST Array 10746311 - -
          Affymetrix Rat Gene 1.0 ST Array 10746303 - -
          Affymetrix Rat Gene 1.0 ST Array 10746318 - -
          Affymetrix Rat Gene 1.0 ST Array 10746324 - -
          Affymetrix Rat Gene 1.0 ST Array 10746315 - -
          Affymetrix Rat Gene 1.0 ST Array 10746316 - -
          Affymetrix Rat Gene 1.0 ST Array 10746307 - -
          Affymetrix Rat Gene 1.0 ST Array 10746317 - -
          Affymetrix Rat Gene 1.1 ST Array 10746293 - -
          Rat Genome 230 2.0 Array 1369698_at - -
          Rat Genome 230 2.0 Array 1369698_PM_at - -
          Affymetrix HT Rat Focus Array 1369698_PM_at - -
          Affymetrix HT Rat Focus Array 1369698_at - -
          Affymetrix Rat Clariom S Assay PSR1000031928.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031897.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031906.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031919.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031893.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031902.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031904.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031905.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031923.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031920.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031925.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031892.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031907.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031916.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031912.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031900.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031910.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031921.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031926.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031915.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031917.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031901.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031924.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031908.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031922.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031909.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031911.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031927.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031914.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031903.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031918.rn.1 TC1000003587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031913.rn.1 TC1000003587.rn.1 RTA TC
          Rat Expression Set 230 1369698_at - -
          Rat Expression Set 230 1369698_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746293 - -
          Rat Genome U34 Set AB010467_s_at - -
          Rat Genome U34 Set AB010467_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20741876 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351428 - -
          RatRef-12 Expression BeadChip ILMN_64863 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline