Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453258_m1
          See other NF2 GE Assays ›
          Gene Symbol:
          Nf2
          Gene Name:
          neurofibromin 2 (merlin)
          Gene Aliases:
          merlin
          Chromosome Location:
          Chr.14: 84996905 - 85088573 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453258_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013193.1 NP_037325.1 2 - 3 642 65
          XM_006251188.2 XP_006251250.1 2 - 3 692 65
          XM_006251191.2 XP_006251253.1 2 - 3 692 65
          GenBank mRNA U61772.1 - 2 - 3 236 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002154.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          negative regulation of protein kinase activity
          ectoderm development
          brain development
          negative regulation of DNA replication
          negative regulation of cell proliferation
          Schwann cell proliferation
          hippocampus development
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          negative regulation of cell growth
          regulation of protein stability
          regulation of hippo signaling
          regulation of cell proliferation
          odontogenesis of dentin-containing tooth
          negative regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of MAPK cascade
          cell-cell junction organization
          positive regulation of cell differentiation
          negative regulation of JAK-STAT cascade
          positive regulation of stress fiber assembly
          lens fiber cell differentiation
          regulation of protein localization to nucleus
          regulation of neural precursor cell proliferation
          actin binding
          integrin binding
          protein binding
          beta-catenin binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14694 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15292 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116814 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116814 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116814 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119413 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P233808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25791 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79717 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85287 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89799 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100870 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778135 - -
          Affymetrix Rat Gene 1.0 ST Array 10778123 - -
          Affymetrix Rat Gene 1.0 ST Array 10778134 - -
          Affymetrix Rat Gene 1.0 ST Array 10778126 - -
          Affymetrix Rat Gene 1.0 ST Array 10778131 - -
          Affymetrix Rat Gene 1.0 ST Array 10778128 - -
          Affymetrix Rat Gene 1.0 ST Array 10778129 - -
          Affymetrix Rat Gene 1.0 ST Array 10778133 - -
          Affymetrix Rat Gene 1.0 ST Array 10778132 - -
          Affymetrix Rat Gene 1.0 ST Array 10778120 - -
          Affymetrix Rat Gene 1.0 ST Array 10778136 - -
          Affymetrix Rat Gene 1.0 ST Array 10778122 - -
          Affymetrix Rat Gene 1.0 ST Array 10778125 - -
          Affymetrix Rat Gene 1.0 ST Array 10778124 - -
          Affymetrix Rat Gene 1.0 ST Array 10778127 - -
          Affymetrix Rat Gene 1.0 ST Array 10778121 - -
          Affymetrix Rat Gene 1.0 ST Array 10778130 - -
          Affymetrix Rat Gene 1.1 ST Array 10778119 - -
          Rat Genome 230 2.0 Array 1396346_at - -
          Rat Genome 230 2.0 Array 1371192_at - -
          Rat Genome 230 2.0 Array 1372502_PM_at - -
          Rat Genome 230 2.0 Array 1372502_at - -
          Rat Genome 230 2.0 Array 1371191_PM_at - -
          Rat Genome 230 2.0 Array 1371191_at - -
          Rat Genome 230 2.0 Array 1371192_PM_at - -
          Rat Genome 230 2.0 Array 1396346_PM_at - -
          Affymetrix HT Rat Focus Array 1371191_PM_at - -
          Affymetrix HT Rat Focus Array 1396346_at - -
          Affymetrix HT Rat Focus Array 1371192_at - -
          Affymetrix HT Rat Focus Array 1371192_PM_at - -
          Affymetrix HT Rat Focus Array 1372502_at - -
          Affymetrix HT Rat Focus Array 1396346_PM_at - -
          Affymetrix HT Rat Focus Array 1371191_at - -
          Affymetrix HT Rat Focus Array 1372502_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400015916.rn.1 TC1400002154.rn.1 RTA TC
          Rat Expression Set 230 1371192_PM_at - -
          Rat Expression Set 230 1372502_at - -
          Rat Expression Set 230 1371192_at - -
          Rat Expression Set 230 1371191_PM_at - -
          Rat Expression Set 230 1371191_at - -
          Rat Expression Set 230 1372502_PM_at - -
          Rat Expression Set 230 1396346_at - -
          Rat Expression Set 230 1396346_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778119 - -
          Rat Genome U34 Set U61772_at - -
          Rat Genome U34 Set rc_AI237525_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21271889 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline