Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453365_m1
          See other RFT1 GE Assays ›
          Gene Symbol:
          Rft1
          Gene Name:
          RFT1 homolog (S. cerevisiae)
          Gene Aliases:
          RGD1562654
          Chromosome Location:
          Chr.16: 6712378 - 6746711 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453365_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rft1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135866.1 NP_001129338.1 11 - 12 1252 65
          XM_006252581.2 XP_006252643.1 11 - 12 1263 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000082.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          glycolipid translocation
          lipid transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P134699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56847 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57177 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134699 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134699 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134699 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17448 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786494 - -
          Affymetrix Rat Gene 1.0 ST Array 10786492 - -
          Affymetrix Rat Gene 1.0 ST Array 10786480 - -
          Affymetrix Rat Gene 1.0 ST Array 10786482 - -
          Affymetrix Rat Gene 1.0 ST Array 10786487 - -
          Affymetrix Rat Gene 1.0 ST Array 10786483 - -
          Affymetrix Rat Gene 1.0 ST Array 10786488 - -
          Affymetrix Rat Gene 1.0 ST Array 10786481 - -
          Affymetrix Rat Gene 1.0 ST Array 10786491 - -
          Affymetrix Rat Gene 1.0 ST Array 10786490 - -
          Affymetrix Rat Gene 1.0 ST Array 10786484 - -
          Affymetrix Rat Gene 1.0 ST Array 10786485 - -
          Affymetrix Rat Gene 1.0 ST Array 10786489 - -
          Affymetrix Rat Gene 1.0 ST Array 10786493 - -
          Affymetrix Rat Gene 1.1 ST Array 10786478 - -
          Rat Genome 230 2.0 Array 1381421_PM_at - -
          Rat Genome 230 2.0 Array 1381421_at - -
          Affymetrix HT Rat Focus Array 1381421_PM_at - -
          Affymetrix HT Rat Focus Array 1381421_at - -
          Affymetrix Rat Clariom S Assay PSR1600000732.rn.1 TC1600000082.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000734.rn.1 TC1600000082.rn.1 RTA TC
          Rat Expression Set 230 1381421_at - -
          Rat Expression Set 230 1381421_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786478 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22025929 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357141 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline