Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453456_g1
          See other TKT GE Assays ›
          Gene Symbol:
          Tkt
          Gene Name:
          transketolase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 6609670 - 6634608 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453456_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tkt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022592.1 NP_072114.1 10 - 11 1374 119
          XM_006252661.1 XP_006252723.1 9 - 10 1406 119
          GenBank mRNA FQ231079.1 - 2 - 3 304 119
          FQ232736.1 - 5 - 6 713 119
          U09256.1 - 10 - 11 1374 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000078.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tkt

          Panther Classification:

          Molecular Function -

          transketolase

          Gene Ontology Categories:

          Function(s) Process(es)

          pentose-phosphate shunt
          metabolic process
          pentose-phosphate shunt, non-oxidative branch
          regulation of growth
          glyceraldehyde-3-phosphate biosynthetic process
          ribose phosphate biosynthetic process
          magnesium ion binding
          transketolase activity
          carbohydrate binding
          thiamine pyrophosphate binding
          protein homodimerization activity
          metal ion binding
          monosaccharide binding
          cofactor binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14221 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14221 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14221 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700625927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63234 - -
          Agilent Rat cDNA Microarray (G4105A) 700625927 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786474 - -
          Affymetrix Rat Gene 1.0 ST Array 10786472 - -
          Affymetrix Rat Gene 1.0 ST Array 10786466 - -
          Affymetrix Rat Gene 1.0 ST Array 10786467 - -
          Affymetrix Rat Gene 1.0 ST Array 10786477 - -
          Affymetrix Rat Gene 1.0 ST Array 10786469 - -
          Affymetrix Rat Gene 1.0 ST Array 10786475 - -
          Affymetrix Rat Gene 1.0 ST Array 10786468 - -
          Affymetrix Rat Gene 1.0 ST Array 10786473 - -
          Affymetrix Rat Gene 1.0 ST Array 10786471 - -
          Affymetrix Rat Gene 1.0 ST Array 10786476 - -
          Affymetrix Rat Gene 1.0 ST Array 10786470 - -
          Affymetrix Rat Gene 1.1 ST Array 10786465 - -
          Rat Genome 230 2.0 Array 1386859_at - -
          Rat Genome 230 2.0 Array 1367611_at - -
          Rat Genome 230 2.0 Array 1376635_at - -
          Rat Genome 230 2.0 Array 1386859_PM_at - -
          Rat Genome 230 2.0 Array 1367611_PM_at - -
          Rat Genome 230 2.0 Array 1376635_PM_at - -
          Affymetrix HT Rat Focus Array 1367611_at - -
          Affymetrix HT Rat Focus Array 1386859_at - -
          Affymetrix HT Rat Focus Array 1386859_PM_at - -
          Affymetrix HT Rat Focus Array 1367611_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600000694.rn.1 TC1600000078.rn.1 RTA TC
          Rat Expression Set 230 1386859_at - -
          Rat Expression Set 230 1367611_PM_at - -
          Rat Expression Set 230 1386859_PM_at - -
          Rat Expression Set 230 1367611_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786465 - -
          Rat Genome U34 Set rc_AI059508_s_at - -
          Rat Genome U34 Set U09256_at - -
          Rat Genome U34 Set rc_AI011684_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21010952 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline