Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453584_m1
          See other PHF7 GE Assays ›
          Gene Symbol:
          Phf7
          Gene Name:
          PHD finger protein 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 7323527 - 7336230 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453584_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012211.1 NP_001012211.1 3 - 4 242 73
          XM_006252632.2 XP_006252694.1 2 - 3 162 73
          GenBank mRNA BC079288.1 - 3 - 4 242 7134345 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001173.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phf7

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1011595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73475 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011595 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011595 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1011595 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011595 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790123 - -
          Affymetrix Rat Gene 1.0 ST Array 10790125 - -
          Affymetrix Rat Gene 1.0 ST Array 10790128 - -
          Affymetrix Rat Gene 1.0 ST Array 10790129 - -
          Affymetrix Rat Gene 1.0 ST Array 10790130 - -
          Affymetrix Rat Gene 1.0 ST Array 10790127 - -
          Affymetrix Rat Gene 1.0 ST Array 10790121 - -
          Affymetrix Rat Gene 1.0 ST Array 10790124 - -
          Affymetrix Rat Gene 1.0 ST Array 10790126 - -
          Affymetrix Rat Gene 1.0 ST Array 10790122 - -
          Affymetrix Rat Gene 1.0 ST Array 10790120 - -
          Affymetrix Rat Gene 1.1 ST Array 10790119 - -
          Rat Genome 230 2.0 Array 1378535_PM_at - -
          Rat Genome 230 2.0 Array 1385808_at - -
          Rat Genome 230 2.0 Array 1378535_at - -
          Rat Genome 230 2.0 Array 1385808_PM_at - -
          Affymetrix HT Rat Focus Array 1385808_at - -
          Affymetrix HT Rat Focus Array 1378535_PM_at - -
          Affymetrix HT Rat Focus Array 1378535_at - -
          Affymetrix HT Rat Focus Array 1385808_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600008843.rn.1 TC1600001173.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600008841.rn.1 TC1600001173.rn.1 RTA TC
          Rat Expression Set 230 1385808_PM_at - -
          Rat Expression Set 230 1385808_at - -
          Rat Expression Set 230 1378535_PM_at - -
          Rat Expression Set 230 1378535_at - -
          Affymetrix Rat Gene 1.0 ST Array 10790119 - -
          Rat Genome U34 Set rc_AI639231_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20870154 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375301 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline