Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453894_m1
          See other RPO1-3 GE Assays ›
          Gene Symbol:
          Rpo1-3
          Gene Name:
          RNA polymerase 1-3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 9693828 - 9727416 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453894_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpo1-3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037787.1 NP_001032876.1 2 - 3 135 155
          GenBank mRNA BC098735.1 - 2 - 3 135 7385125 155
          FQ221941.1 - 2 - 3 212 155
          FQ232634.1 - 2 - 3 194 155

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001052.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpo1-3

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase I promoter
          transcription from RNA polymerase III promoter
          RNA polymerase I activity
          RNA polymerase III activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P187238 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85204 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58630 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756496 - -
          Affymetrix Rat Gene 1.0 ST Array 10756493 - -
          Affymetrix Rat Gene 1.0 ST Array 10756495 - -
          Affymetrix Rat Gene 1.1 ST Array 10756492 - -
          Rat Genome 230 2.0 Array 1388404_at - -
          Rat Genome 230 2.0 Array 1371351_at - -
          Rat Genome 230 2.0 Array 1371351_PM_at - -
          Rat Genome 230 2.0 Array 1388404_PM_at - -
          Affymetrix HT Rat Focus Array 1371351_at - -
          Affymetrix HT Rat Focus Array 1388404_at - -
          Affymetrix HT Rat Focus Array 1371351_PM_at - -
          Affymetrix HT Rat Focus Array 1388404_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200008868.rn.1 TC1200001052.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008869.rn.1 TC1200001052.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008871.rn.1 TC1200001052.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008876.rn.1 TC1200001052.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008865.rn.1 TC1200001052.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008867.rn.1 TC1200001052.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008866.rn.1 TC1200001052.rn.1 RTA TC
          Rat Expression Set 230 1371351_PM_at - -
          Rat Expression Set 230 1388404_PM_at - -
          Rat Expression Set 230 1388404_at - -
          Rat Expression Set 230 1371351_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756492 - -
          Rat Genome U34 Set rc_AA799724_at - -
          Rat Genome U34 Set rc_AA799724_g_at - -
          Rat Genome U34 Set rc_AI169166_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20841547 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362729 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline