Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453962_m1
          See other UROS GE Assays ›
          Gene Symbol:
          Uros
          Gene Name:
          uroporphyrinogen III synthase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 205755766 - 205777167 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453962_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uros
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012068.1 NP_001012068.1 9 - 10 819 70
          XM_008759890.1 XP_008758112.1 8 - 9 1020 70
          XM_008759891.1 XP_008758113.1 8 - 9 715 70
          XM_008759892.1 XP_008758114.1 8 - 9 988 70
          GenBank mRNA BC083730.1 - 9 - 10 819 7191339 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006816.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uros

          Panther Classification:

          Molecular Function -

          hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          porphyrin-containing compound biosynthetic process
          uroporphyrinogen III biosynthetic process
          heme biosynthetic process
          response to antibiotic
          cellular response to arsenic-containing substance
          cellular response to amine stimulus
          uroporphyrinogen-III synthase activity
          cofactor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1034637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163730 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034637 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034637 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1034637 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100197 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726352 - -
          Affymetrix Rat Gene 1.0 ST Array 10726355 - -
          Affymetrix Rat Gene 1.0 ST Array 10726353 - -
          Affymetrix Rat Gene 1.0 ST Array 10726351 - -
          Affymetrix Rat Gene 1.0 ST Array 10726347 - -
          Affymetrix Rat Gene 1.0 ST Array 10726356 - -
          Affymetrix Rat Gene 1.0 ST Array 10726349 - -
          Affymetrix Rat Gene 1.0 ST Array 10726350 - -
          Affymetrix Rat Gene 1.0 ST Array 10726357 - -
          Affymetrix Rat Gene 1.0 ST Array 10726348 - -
          Affymetrix Rat Gene 1.0 ST Array 10726354 - -
          Affymetrix Rat Gene 1.1 ST Array 10726346 - -
          Rat Genome 230 2.0 Array 1392695_a_at - -
          Rat Genome 230 2.0 Array 1392695_PM_a_at - -
          Affymetrix HT Rat Focus Array 1392695_a_at - -
          Affymetrix HT Rat Focus Array 1392695_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100053705.rn.1 TC0100006816.rn.1 RTA TC
          Rat Expression Set 230 1392695_a_at - -
          Rat Expression Set 230 1392695_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726346 - -
          Rat Genome U34 Set rc_AA900913_g_at - -
          Rat Genome U34 Set rc_AA900913_at - -
          Rat Genome U34 Set rc_AI178319_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20975044 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376774 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline