Search
Search

Ehbp1l1| Gene Symbol: Ehbp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025117.2 | NP_001020288.2 | 12 - 13 | 1950 | 60 | |
| NM_001129997.1 | NP_001123469.1 | 12 - 13 | 4722 | 60 | ||
| NM_001270704.1 | NP_001257633.1 | 12 - 13 | 2445 | 60 | ||
| NM_001270705.1 | NP_001257634.1 | 11 - 12 | 1914 | 60 | ||
| XM_008760139.1 | XP_008758361.1 | 12 - 13 | 2480 | 60 | ||
| GenBank mRNA | BC087115.1 | - | 11 - 12 | 1877 | 60 | |
| BC129126.1 | - | 12 - 13 | 1950 | 60 | ||
| BC167745.1 | - | 12 - 13 | 2226 | 7310903 | 60 | |
Set Membership: |
|