Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454057_m1
          See other METTL10 GE Assays ›
          Gene Symbol:
          Mettl10
          Gene Name:
          methyltransferase like 10
          Gene Aliases:
          RGD1306300
          Chromosome Location:
          Chr.1: 204829977 - 204855627 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454057_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mettl10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108504.1 NP_001101974.1 5 - 6 687 68
          XM_006230441.1 XP_006230503.1 5 - 6 682 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006799.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          methylation
          methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134687 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17077 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17077 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17077 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726312 - -
          Affymetrix Rat Gene 1.0 ST Array 10726314 - -
          Affymetrix Rat Gene 1.0 ST Array 10726310 - -
          Affymetrix Rat Gene 1.0 ST Array 10726309 - -
          Affymetrix Rat Gene 1.0 ST Array 10726313 - -
          Affymetrix Rat Gene 1.0 ST Array 10726311 - -
          Affymetrix Rat Gene 1.1 ST Array 10726308 - -
          Rat Genome 230 2.0 Array 1382290_at - -
          Rat Genome 230 2.0 Array 1382290_PM_at - -
          Affymetrix HT Rat Focus Array 1382290_at - -
          Affymetrix HT Rat Focus Array 1382290_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100053614.rn.1 TC0100006799.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100053615.rn.1 TC0100006799.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100053616.rn.1 TC0100006799.rn.1 RTA TC
          Rat Expression Set 230 1382290_PM_at - -
          Rat Expression Set 230 1382290_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726308 - -
          Rat Genome U34 Set rc_AI070033_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22127476 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367272 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline