Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454136_g1
          See other SIPA1 GE Assays ›
          Gene Symbol:
          Sipa1
          Gene Name:
          signal-induced proliferation-associated 1
          Gene Aliases:
          Spa1
          Chromosome Location:
          Chr.1: 221006301 - 221018770 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454136_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sipa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004089.1 NP_001004089.1 14 - 15 2981 83
          XM_006230849.2 XP_006230911.1 15 - 16 3598 83
          GenBank mRNA AB091474.1 - 14 - 15 2981 83
          FQ223350.1 - 15 - 16 3188 83
          FQ228273.1 - 15 - 16 3167 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sipa1

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to water deprivation
          positive regulation of GTPase activity
          regulation of small GTPase mediated signal transduction
          regulation of cell cycle
          GTPase activator activity
          protein C-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P433259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P433259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P433259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P433259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727937 - -
          Affymetrix Rat Gene 1.0 ST Array 10727940 - -
          Affymetrix Rat Gene 1.0 ST Array 10727930 - -
          Affymetrix Rat Gene 1.0 ST Array 10727936 - -
          Affymetrix Rat Gene 1.0 ST Array 10727931 - -
          Affymetrix Rat Gene 1.0 ST Array 10727934 - -
          Affymetrix Rat Gene 1.0 ST Array 10727935 - -
          Affymetrix Rat Gene 1.0 ST Array 10727928 - -
          Affymetrix Rat Gene 1.0 ST Array 10727939 - -
          Affymetrix Rat Gene 1.0 ST Array 10727933 - -
          Affymetrix Rat Gene 1.0 ST Array 10727938 - -
          Affymetrix Rat Gene 1.0 ST Array 10727932 - -
          Affymetrix Rat Gene 1.0 ST Array 10727927 - -
          Affymetrix Rat Gene 1.0 ST Array 10727929 - -
          Affymetrix Rat Gene 1.0 ST Array 10727926 - -
          Affymetrix Rat Gene 1.1 ST Array 10727924 - -
          Rat Genome 230 2.0 Array 1375709_PM_at - -
          Rat Genome 230 2.0 Array 1375709_at - -
          Affymetrix HT Rat Focus Array 1375709_PM_at - -
          Affymetrix HT Rat Focus Array 1375709_at - -
          Affymetrix Rat Clariom S Assay PSR0100057007.rn.1 TC0100007128.rn.1 RTA TC
          Rat Expression Set 230 1375709_PM_at - -
          Rat Expression Set 230 1375709_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727924 - -
          Rat Genome U34 Set rc_AI180414_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21215045 - -
          ABI Rat Genome Survey Microarray 21862905 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368126 - -
          RatRef-12 Expression BeadChip ILMN_51994 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline