Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454518_m1
          See other IGF2 GE Assays ›
          Gene Symbol:
          Igf2
          Gene Name:
          insulin-like growth factor 2
          Gene Aliases:
          IGFII, RNIGF2
          Chromosome Location:
          Chr.1: 215828102 - 215839081 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454518_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190162.1 NP_001177091.1 2 - 3 243 74
          NM_001190163.1 NP_001177092.1 2 - 3 231 74
          NM_031511.2 NP_113699.2 2 - 3 1218 74
          XM_006230664.2 XP_006230726.1 2 - 3 1020 74
          XM_006230665.2 XP_006230727.1 2 - 3 223 74
          XM_008760074.1 XP_008758296.1 2 - 3 1027 74
          XM_008760075.1 XP_008758297.1 3 - 4 556 74
          GenBank mRNA JQ408382.1 - 1156 74
          X00911.1 - 2 - 3 1218 74
          X13101.1 - 2 - 3 255 74
          X14833.1 - 2 - 3 531 74
          X14834.1 - 2 - 3 1286 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006984.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igf2

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          positive regulation of protein phosphorylation
          glucose metabolic process
          female pregnancy
          memory
          cell proliferation
          response to radiation
          organ morphogenesis
          response to organic cyclic compound
          exocrine pancreas development
          response to nutrient levels
          response to estradiol
          response to nicotine
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          wound healing
          positive regulation of activated T cell proliferation
          response to drug
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          response to ethanol
          positive regulation of glycogen biosynthetic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of insulin receptor signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          striated muscle cell differentiation
          positive regulation of protein kinase B signaling
          cellular response to mechanical stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of steroid hormone biosynthetic process
          positive regulation of receptor activity
          positive regulation of glycogen (starch) synthase activity
          insulin receptor binding
          insulin-like growth factor receptor binding
          hormone activity
          protein binding
          growth factor activity
          receptor activator activity
          protein serine/threonine kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1037953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238374 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27735 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037953 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037953 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1037953 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1037953 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702024981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90118 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37463 - -
          Agilent Rat cDNA Microarray (G4105A) 702024981 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727002 - -
          Affymetrix Rat Gene 1.0 ST Array 10727005 - -
          Affymetrix Rat Gene 1.0 ST Array 10727000 - -
          Affymetrix Rat Gene 1.0 ST Array 10727004 - -
          Affymetrix Rat Gene 1.0 ST Array 10727001 - -
          Affymetrix Rat Gene 1.1 ST Array 10726999 - -
          Rat Genome 230 2.0 Array 1367571_a_at - -
          Rat Genome 230 2.0 Array 1398322_at - -
          Rat Genome 230 2.0 Array 1367571_PM_a_at - -
          Rat Genome 230 2.0 Array 1398322_PM_at - -
          Rat Genome 230 2.0 Array 1388252_PM_at - -
          Rat Genome 230 2.0 Array 1371206_PM_a_at - -
          Rat Genome 230 2.0 Array 1388252_at - -
          Rat Genome 230 2.0 Array 1371206_a_at - -
          Affymetrix HT Rat Focus Array 1388252_at - -
          Affymetrix HT Rat Focus Array 1367571_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388252_PM_at - -
          Affymetrix HT Rat Focus Array 1398322_at - -
          Affymetrix HT Rat Focus Array 1398322_PM_at - -
          Affymetrix HT Rat Focus Array 1371206_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371206_a_at - -
          Affymetrix HT Rat Focus Array 1367571_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100055209.rn.1 TC0100006984.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055218.rn.1 TC0100006984.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055205.rn.1 TC0100006984.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055208.rn.1 TC0100006984.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055206.rn.1 TC0100006984.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055212.rn.1 TC0100006984.rn.1 RTA TC
          Rat Expression Set 230 1371206_a_at - -
          Rat Expression Set 230 1371206_PM_a_at - -
          Rat Expression Set 230 1398322_at - -
          Rat Expression Set 230 1388252_PM_at - -
          Rat Expression Set 230 1367571_PM_a_at - -
          Rat Expression Set 230 1367571_a_at - -
          Rat Expression Set 230 1398322_PM_at - -
          Rat Expression Set 230 1388252_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726999 - -
          Rat Genome U34 Set M17960_at - -
          Rat Genome U34 Set E00988mRNA_s_at - -
          Rat Genome U34 Set X16703_r_at - -
          Rat Genome U34 Set X16703_i_at - -
          Rat Genome U34 Set X17012mRNA_s_at - -
          Rat Genome U34 Set M17960_g_at - -
          Rat Genome U34 Set X16703_i_at - -
          Rat Genome U34 Set X16703_r_at - -
          Rat Genome U34 Set M17960_at - -
          Rat Genome U34 Set X17012mRNA_s_at - -
          Rat Genome U34 Set E00988mRNA_s_at - -
          Rat Genome U34 Set M17960_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21268639 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359301 - -
          RatRef-12 Expression BeadChip ILMN_63177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline