Search
Search
| Gene Symbol: Myrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170487.1 | NP_001163958.1 | 14 - 15 | 2007 | 81 | |
| XM_006231030.2 | XP_006231092.1 | 14 - 15 | 2168 | 81 | ||
| XM_006231031.2 | XP_006231093.1 | 14 - 15 | 2168 | 81 | ||
| XM_008760220.1 | XP_008758442.1 | 14 - 15 | 2168 | 81 | ||
| XM_008760221.1 | XP_008758443.1 | 13 - 14 | 1904 | 81 | ||
| XM_008760222.1 | XP_008758444.1 | 11 - 12 | 1728 | 81 | ||
Set Membership: |
|