Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454738_m1
          See other CYP2G1 GE Assays ›
          Gene Symbol:
          Cyp2g1
          Gene Name:
          cytochrome P450, family 2, subfamily g, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 83626639 - 83637780 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454738_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2g1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012787.1 NP_036919.1 8 - 9 1303 86
          GenBank mRNA M33296.2 - 8 - 9 1301 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001102.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          sensory perception of smell
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P391192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P391192 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P391192 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P391192 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391192 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705272 - -
          Affymetrix Rat Gene 1.0 ST Array 10705268 - -
          Affymetrix Rat Gene 1.0 ST Array 10705269 - -
          Affymetrix Rat Gene 1.0 ST Array 10705275 - -
          Affymetrix Rat Gene 1.0 ST Array 10705273 - -
          Affymetrix Rat Gene 1.0 ST Array 10705274 - -
          Affymetrix Rat Gene 1.0 ST Array 10705267 - -
          Affymetrix Rat Gene 1.0 ST Array 10705270 - -
          Affymetrix Rat Gene 1.0 ST Array 10705271 - -
          Affymetrix Rat Gene 1.1 ST Array 10705266 - -
          Rat Genome 230 2.0 Array 1371142_PM_at - -
          Rat Genome 230 2.0 Array 1371142_at - -
          Affymetrix HT Rat Focus Array 1371142_at - -
          Affymetrix HT Rat Focus Array 1371142_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100008032.rn.1 TC0100001102.rn.1 RTA TC
          Rat Expression Set 230 1371142_at - -
          Rat Expression Set 230 1371142_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705266 - -
          Rat Genome U34 Set M31931cds_s_at - -
          Rat Genome U34 Set M31931cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21322876 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354124 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline