Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455333_m1
          See other GATAD2B GE Assays ›
          Gene Symbol:
          Gatad2b
          Gene Name:
          GATA zinc finger domain containing 2B
          Gene Aliases:
          RGD1308533
          Chromosome Location:
          Chr.2: 189654874 - 189735056 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455333_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gatad2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024888.1 NP_001020059.1 10 - 11 1848 82
          XM_008761144.1 XP_008759366.1 10 - 11 1779 82
          XM_008761145.1 XP_008759367.1 10 - 11 1776 82
          GenBank mRNA BC097428.1 - 10 - 11 1848 7455294 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001607.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gatad2b

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          ATP-dependent chromatin remodeling
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          zinc ion binding
          nucleosomal DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P516658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47348 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35534 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P516658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P516658 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P516658 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8196 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89481 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3880 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817018 - -
          Affymetrix Rat Gene 1.0 ST Array 10817011 - -
          Affymetrix Rat Gene 1.0 ST Array 10817019 - -
          Affymetrix Rat Gene 1.0 ST Array 10817020 - -
          Affymetrix Rat Gene 1.0 ST Array 10817014 - -
          Affymetrix Rat Gene 1.0 ST Array 10817017 - -
          Affymetrix Rat Gene 1.0 ST Array 10817016 - -
          Affymetrix Rat Gene 1.0 ST Array 10817012 - -
          Affymetrix Rat Gene 1.0 ST Array 10817015 - -
          Affymetrix Rat Gene 1.0 ST Array 10817013 - -
          Affymetrix Rat Gene 1.1 ST Array 10817010 - -
          Rat Genome 230 2.0 Array 1392552_PM_at - -
          Rat Genome 230 2.0 Array 1392552_at - -
          Affymetrix HT Rat Focus Array 1392552_PM_at - -
          Affymetrix HT Rat Focus Array 1392552_at - -
          Affymetrix Rat Clariom S Assay PSR0200010754.rn.1 TC0200001607.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010755.rn.1 TC0200001607.rn.1 RTA TC
          Rat Expression Set 230 1392552_at - -
          Rat Expression Set 230 1392552_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817010 - -
          Rat Genome U34 Set rc_AI045118_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21561848 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358423 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline