Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455421_m1
          See other ZBBX GE Assays ›
          Gene Symbol:
          Zbbx
          Gene Name:
          zinc finger, B-box domain containing
          Gene Aliases:
          RGD1308632
          Chromosome Location:
          Chr.2: 173436058 - 173551721 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455421_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbbx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029922.1 NP_001025093.1 13 - 14 1492 135
          XM_006232495.2 XP_006232557.1 13 - 14 1423 135
          XM_006232496.2 XP_006232558.1 13 - 14 1422 135
          XM_008761103.1 XP_008759325.1 15 - 16 1870 135
          XM_008761104.1 XP_008759326.1 15 - 16 1854 135
          XM_008761105.1 XP_008759327.1 15 - 16 1873 135
          GenBank mRNA BC097393.1 - 13 - 14 1492 7453249 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003885.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbbx

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157652 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41930 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039091 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52531 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69226 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823687 - -
          Affymetrix Rat Gene 1.0 ST Array 10823694 - -
          Affymetrix Rat Gene 1.0 ST Array 10823700 - -
          Affymetrix Rat Gene 1.0 ST Array 10823693 - -
          Affymetrix Rat Gene 1.0 ST Array 10823691 - -
          Affymetrix Rat Gene 1.0 ST Array 10823701 - -
          Affymetrix Rat Gene 1.0 ST Array 10823698 - -
          Affymetrix Rat Gene 1.0 ST Array 10823696 - -
          Affymetrix Rat Gene 1.0 ST Array 10823692 - -
          Affymetrix Rat Gene 1.0 ST Array 10823697 - -
          Affymetrix Rat Gene 1.0 ST Array 10823699 - -
          Affymetrix Rat Gene 1.0 ST Array 10823688 - -
          Affymetrix Rat Gene 1.0 ST Array 10823689 - -
          Affymetrix Rat Gene 1.0 ST Array 10823702 - -
          Affymetrix Rat Gene 1.0 ST Array 10823695 - -
          Affymetrix Rat Gene 1.1 ST Array 10823683 - -
          Affymetrix Rat Clariom S Assay PSR0200026000.rn.1 TC0200003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026002.rn.1 TC0200003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026003.rn.1 TC0200003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026004.rn.1 TC0200003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026001.rn.1 TC0200003885.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10823683 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21431715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364582 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline