Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455453_m1
          See other GON4L GE Assays ›
          Gene Symbol:
          Gon4l
          Gene Name:
          gon-4-like (C. elegans)
          Gene Aliases:
          Dingo, Yarp
          Chromosome Location:
          Chr.2: 188139730 - 188212902 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455453_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gon4l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024797.1 NP_001019968.1 29 - 30 5953 101
          XM_006232735.2 XP_006232797.1 29 - 30 5944 101
          XM_006232736.2 XP_006232798.1 29 - 30 5929 101
          XM_006232737.2 XP_006232799.1 28 - 29 5908 101
          XM_006232738.2 XP_006232800.1 28 - 29 5905 101
          XM_006232739.2 XP_006232801.1 29 - 30 5884 101
          XM_006232740.2 XP_006232802.1 28 - 29 5740 101
          XM_006232741.2 XP_006232803.1 28 - 29 5734 101
          XM_008761201.1 XP_008759423.1 30 - 31 5958 101
          XM_008761202.1 XP_008759424.1 28 - 29 5393 101
          GenBank mRNA AB195689.1 - 28 - 29 5787 101
          AY724781.1 - 29 - 30 5953 101
          AY724782.1 - 28 - 29 6033 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004976.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gon4l

          Panther Classification:

          Molecular Function -

          transcription cofactor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          B cell differentiation
          DNA binding
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P349408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P349408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P349408 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P349408 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P349408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95624 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97514 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816631 - -
          Affymetrix Rat Gene 1.0 ST Array 10816624 - -
          Affymetrix Rat Gene 1.0 ST Array 10816639 - -
          Affymetrix Rat Gene 1.0 ST Array 10816643 - -
          Affymetrix Rat Gene 1.0 ST Array 10816646 - -
          Affymetrix Rat Gene 1.0 ST Array 10816625 - -
          Affymetrix Rat Gene 1.0 ST Array 10816621 - -
          Affymetrix Rat Gene 1.0 ST Array 10816619 - -
          Affymetrix Rat Gene 1.0 ST Array 10816617 - -
          Affymetrix Rat Gene 1.0 ST Array 10816637 - -
          Affymetrix Rat Gene 1.0 ST Array 10816629 - -
          Affymetrix Rat Gene 1.0 ST Array 10816626 - -
          Affymetrix Rat Gene 1.0 ST Array 10816616 - -
          Affymetrix Rat Gene 1.0 ST Array 10816614 - -
          Affymetrix Rat Gene 1.0 ST Array 10816630 - -
          Affymetrix Rat Gene 1.0 ST Array 10816638 - -
          Affymetrix Rat Gene 1.0 ST Array 10816627 - -
          Affymetrix Rat Gene 1.0 ST Array 10816623 - -
          Affymetrix Rat Gene 1.0 ST Array 10816622 - -
          Affymetrix Rat Gene 1.0 ST Array 10816615 - -
          Affymetrix Rat Gene 1.0 ST Array 10816628 - -
          Affymetrix Rat Gene 1.0 ST Array 10816642 - -
          Affymetrix Rat Gene 1.0 ST Array 10816620 - -
          Affymetrix Rat Gene 1.0 ST Array 10816635 - -
          Affymetrix Rat Gene 1.0 ST Array 10816640 - -
          Affymetrix Rat Gene 1.0 ST Array 10816644 - -
          Affymetrix Rat Gene 1.0 ST Array 10816645 - -
          Affymetrix Rat Gene 1.0 ST Array 10816632 - -
          Affymetrix Rat Gene 1.0 ST Array 10816618 - -
          Affymetrix Rat Gene 1.0 ST Array 10816636 - -
          Affymetrix Rat Gene 1.1 ST Array 10824357 - -
          Affymetrix Rat Gene 1.1 ST Array 10816613 - -
          Affymetrix Rat Clariom S Assay PSR0200034647.rn.1 TC0200004976.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034648.rn.1 TC0200004976.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10816613 - -
          Rat Genome U34 Set rc_AI030302_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21738646 - -
          ABI Rat Genome Survey Microarray 21859259 - -
          ABI Rat Genome Survey Microarray 22120085 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649966 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline