Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455552_m1
          See other RBM46 GE Assays ›
          Gene Symbol:
          Rbm46
          Gene Name:
          RNA binding motif protein 46
          Gene Aliases:
          RGD1560205
          Chromosome Location:
          Chr.2: 181828932 - 181874223 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455552_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135717.1 NP_001129189.1 2 - 3 334 128
          XM_006232530.2 XP_006232592.1 2 - 3 329 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003937.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          nucleic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P426660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P426660 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P426660 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P426660 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P426660 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823944 - -
          Affymetrix Rat Gene 1.0 ST Array 10823948 - -
          Affymetrix Rat Gene 1.0 ST Array 10823946 - -
          Affymetrix Rat Gene 1.0 ST Array 10823945 - -
          Affymetrix Rat Gene 1.0 ST Array 10823947 - -
          Affymetrix Rat Gene 1.0 ST Array 10823942 - -
          Affymetrix Rat Gene 1.1 ST Array 10823941 - -
          Affymetrix Rat Clariom S Assay PSR0200026447.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026448.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026450.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026457.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026452.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026440.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026449.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026456.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026451.rn.1 TC0200003937.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10823941 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20771778 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368556 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline