Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455817_m1
          See other C1GALT1 GE Assays ›
          Gene Symbol:
          C1galt1
          Gene Name:
          core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 33890089 - 33943219 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455817_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C1galt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022950.1 NP_075239.1 3 - 4 1037 124
          XM_006236091.2 XP_006236153.1 4 - 5 1058 124
          XM_008762725.1 XP_008760947.1 4 - 5 1007 124
          GenBank mRNA AF157963.1 - 3 - 4 1037 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000349.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C1galt1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          kidney development
          protein O-linked glycosylation
          O-glycan processing
          intestinal epithelial cell development
          glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P187195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P187195 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P187195 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P187195 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P187195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15034 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37180 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853695 - -
          Affymetrix Rat Gene 1.0 ST Array 10853694 - -
          Affymetrix Rat Gene 1.0 ST Array 10853692 - -
          Affymetrix Rat Gene 1.0 ST Array 10853693 - -
          Affymetrix Rat Gene 1.1 ST Array 10853691 - -
          Rat Genome 230 2.0 Array 1387446_at - -
          Rat Genome 230 2.0 Array 1387446_PM_at - -
          Affymetrix HT Rat Focus Array 1387446_PM_at - -
          Affymetrix HT Rat Focus Array 1387446_at - -
          Affymetrix Rat Clariom S Assay PSR0400002627.rn.1 TC0400000349.rn.1 RTA TC
          Rat Expression Set 230 1387446_PM_at - -
          Rat Expression Set 230 1387446_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853691 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20742628 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362982 - -
          RatRef-12 Expression BeadChip ILMN_66836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline