Search
Search
| Gene Symbol: Cep112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105849.1 | NP_001099319.1 | 4 - 5 | 636 | 61 | |
| XM_008768317.1 | XP_008766539.1 | 4 - 5 | 670 | 61 | ||
| XM_008768318.1 | XP_008766540.1 | 4 - 5 | 808 | 61 | ||
| XM_008768319.1 | XP_008766541.1 | 4 - 5 | 670 | 61 | ||
| XM_008768320.1 | XP_008766542.1 | 4 - 5 | 750 | 61 | ||
| XM_008768321.1 | XP_008766543.1 | 4 - 5 | 1244 | 61 | ||
Set Membership: |
|