Search
Search
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106990.1 | NP_001100460.1 | 5 - 6 | 2418 | 99 | |
| XM_008767570.1 | XP_008765792.1 | 5 - 6 | 9837 | 99 | ||
| XM_008767571.1 | XP_008765793.1 | 6 - 7 | 2951 | 99 | ||
| XM_008767572.1 | XP_008765794.1 | 5 - 6 | 9837 | 99 | ||
| XM_008767573.1 | XP_008765795.1 | 5 - 6 | 9837 | 99 | ||
| XM_008767574.1 | XP_008765796.1 | 5 - 6 | 9837 | 99 | ||
| XM_008767575.1 | XP_008765797.1 | 5 - 6 | 9837 | 99 | ||
| XM_008767576.1 | XP_008765798.1 | 5 - 6 | 2661 | 99 | ||
Set Membership: |
|