Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01456616_m1
          See other MRC2 GE Assays ›
          Gene Symbol:
          Mrc2
          Gene Name:
          mannose receptor, C type 2
          Gene Aliases:
          Endo180
          Chromosome Location:
          Chr.10: 93520311 - 93581599 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01456616_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024687.1 NP_001019858.1 20 - 21 2947 110
          XM_006247631.2 XP_006247693.1 20 - 21 3072 110
          GenBank mRNA DQ058624.1 - 20 - 21 2947 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001755.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrc2

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          endocytosis
          collagen catabolic process
          collagen binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P122474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89480 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96153 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101276 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122474 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P122474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92112 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28690 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739000 - -
          Affymetrix Rat Gene 1.0 ST Array 10738988 - -
          Affymetrix Rat Gene 1.0 ST Array 10738973 - -
          Affymetrix Rat Gene 1.0 ST Array 10738999 - -
          Affymetrix Rat Gene 1.0 ST Array 10738991 - -
          Affymetrix Rat Gene 1.0 ST Array 10738992 - -
          Affymetrix Rat Gene 1.0 ST Array 10738990 - -
          Affymetrix Rat Gene 1.0 ST Array 10738974 - -
          Affymetrix Rat Gene 1.0 ST Array 10738998 - -
          Affymetrix Rat Gene 1.0 ST Array 10738996 - -
          Affymetrix Rat Gene 1.0 ST Array 10738987 - -
          Affymetrix Rat Gene 1.0 ST Array 10738985 - -
          Affymetrix Rat Gene 1.0 ST Array 10738979 - -
          Affymetrix Rat Gene 1.0 ST Array 10738976 - -
          Affymetrix Rat Gene 1.0 ST Array 10738978 - -
          Affymetrix Rat Gene 1.0 ST Array 10739001 - -
          Affymetrix Rat Gene 1.0 ST Array 10738977 - -
          Affymetrix Rat Gene 1.0 ST Array 10738986 - -
          Affymetrix Rat Gene 1.0 ST Array 10738984 - -
          Affymetrix Rat Gene 1.0 ST Array 10738993 - -
          Affymetrix Rat Gene 1.0 ST Array 10738982 - -
          Affymetrix Rat Gene 1.0 ST Array 10738997 - -
          Affymetrix Rat Gene 1.0 ST Array 10738975 - -
          Affymetrix Rat Gene 1.0 ST Array 10738995 - -
          Affymetrix Rat Gene 1.0 ST Array 10738980 - -
          Affymetrix Rat Gene 1.0 ST Array 10738983 - -
          Affymetrix Rat Gene 1.0 ST Array 10738989 - -
          Affymetrix Rat Gene 1.0 ST Array 10738994 - -
          Affymetrix Rat Gene 1.0 ST Array 10738981 - -
          Affymetrix Rat Gene 1.1 ST Array 10738972 - -
          Affymetrix Rat Clariom S Assay PSR1000016078.rn.1 TC1000001755.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10738972 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21338402 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline