Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01456903_m1
          See other GNPTG GE Assays ›
          Gene Symbol:
          Gnptg
          Gene Name:
          N-acetylglucosamine-1-phosphate transferase, gamma subunit
          Gene Aliases:
          Gnptag
          Chromosome Location:
          Chr.10: 14593050 - 14597986 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01456903_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnptg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100493.1 NP_001093963.1 3 - 4 198 69
          XM_006245935.2 XP_006245997.1 3 - 4 215 69
          GenBank mRNA BC098716.1 - 3 - 4 180 69
          BC158818.1 - 3 - 4 198 7374209 69
          FQ213320.1 - 3 - 4 205 69
          FQ213586.1 - 3 - 4 163 69
          FQ213702.1 - 3 - 4 197 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002420.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          N-glycan processing to lysosome
          carbohydrate phosphorylation
          glycoprotein binding
          UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosaminephosphotransferase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98521 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148762 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148762 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148762 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128912 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61302 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741370 - -
          Affymetrix Rat Gene 1.0 ST Array 10741365 - -
          Affymetrix Rat Gene 1.0 ST Array 10741369 - -
          Affymetrix Rat Gene 1.0 ST Array 10741362 - -
          Affymetrix Rat Gene 1.0 ST Array 10741371 - -
          Affymetrix Rat Gene 1.0 ST Array 10741372 - -
          Affymetrix Rat Gene 1.0 ST Array 10741368 - -
          Affymetrix Rat Gene 1.0 ST Array 10741364 - -
          Affymetrix Rat Gene 1.0 ST Array 10741366 - -
          Affymetrix Rat Gene 1.0 ST Array 10741367 - -
          Affymetrix Rat Gene 1.0 ST Array 10741363 - -
          Affymetrix Rat Gene 1.1 ST Array 10741361 - -
          Rat Genome 230 2.0 Array 1376492_PM_at - -
          Rat Genome 230 2.0 Array 1386622_at - -
          Rat Genome 230 2.0 Array 1374790_at - -
          Rat Genome 230 2.0 Array 1376492_at - -
          Rat Genome 230 2.0 Array 1374790_PM_at - -
          Rat Genome 230 2.0 Array 1386622_PM_at - -
          Affymetrix HT Rat Focus Array 1374790_at - -
          Affymetrix HT Rat Focus Array 1374790_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000021700.rn.1 TC1000002420.rn.1 RTA TC
          Rat Expression Set 230 1374790_PM_at - -
          Rat Expression Set 230 1374790_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741361 - -
          Rat Genome U34 Set rc_AA875171_at - -
          Rat Genome U34 Set rc_AA875171_g_at - -
          Rat Genome U34 Set rc_AA850288_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21770151 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358439 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline