Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457219_m1
          See other MMD2 GE Assays ›
          Gene Symbol:
          Mmd2
          Gene Name:
          monocyte to macrophage differentiation-associated 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 14021724 - 14078121 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457219_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037217.1 NP_001032294.1 6 - 7 714 59
          GenBank mRNA BC161816.1 - 6 - 7 692 7320318 59
          CO404759.1 - 3 - 4 254 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000254.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmd2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s)

          cytolysis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70587 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095005 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095005 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095005 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217648 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36878 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756846 - -
          Affymetrix Rat Gene 1.0 ST Array 10756848 - -
          Affymetrix Rat Gene 1.0 ST Array 10756849 - -
          Affymetrix Rat Gene 1.0 ST Array 10756851 - -
          Affymetrix Rat Gene 1.0 ST Array 10756845 - -
          Affymetrix Rat Gene 1.0 ST Array 10756847 - -
          Affymetrix Rat Gene 1.0 ST Array 10756852 - -
          Affymetrix Rat Gene 1.0 ST Array 10756850 - -
          Affymetrix Rat Gene 1.1 ST Array 10756844 - -
          Rat Genome 230 2.0 Array 1389221_at - -
          Rat Genome 230 2.0 Array 1389221_PM_at - -
          Affymetrix HT Rat Focus Array 1389221_at - -
          Affymetrix HT Rat Focus Array 1389221_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001864.rn.1 TC1200000254.rn.1 RTA TC
          Rat Expression Set 230 1389221_PM_at - -
          Rat Expression Set 230 1389221_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756844 - -
          Rat Genome U34 Set rc_AI136114_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20961623 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356032 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline