Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457237_m1
          See other CCZ1B GE Assays ›
          Gene Symbol:
          Ccz1b
          Gene Name:
          CCZ1 vacuolar protein trafficking and biogenesis associated
          Gene Aliases:
          Ccz1, RGD1306410
          Chromosome Location:
          Chr.12: 12638854 - 12662371 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457237_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccz1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014126.1 NP_001014148.1 13 - 14 2346 63
          XM_006248878.1 XP_006248940.1 14 - 15 1469 63
          XM_006248879.1 XP_006248941.1 13 - 14 1310 63
          GenBank mRNA BC079086.1 - 13 - 14 2346 7123188 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000215.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccz1b

          Gene Ontology Categories:

          Function(s)

          vesicle-mediated transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060028 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P339881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81652 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78923 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P339881 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P339881 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P339881 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P339881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3374 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104069 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756674 - -
          Affymetrix Rat Gene 1.0 ST Array 10756682 - -
          Affymetrix Rat Gene 1.0 ST Array 10756685 - -
          Affymetrix Rat Gene 1.0 ST Array 10756675 - -
          Affymetrix Rat Gene 1.0 ST Array 10756677 - -
          Affymetrix Rat Gene 1.0 ST Array 10756676 - -
          Affymetrix Rat Gene 1.0 ST Array 10756681 - -
          Affymetrix Rat Gene 1.0 ST Array 10756688 - -
          Affymetrix Rat Gene 1.0 ST Array 10756683 - -
          Affymetrix Rat Gene 1.0 ST Array 10756689 - -
          Affymetrix Rat Gene 1.0 ST Array 10756678 - -
          Affymetrix Rat Gene 1.0 ST Array 10756684 - -
          Affymetrix Rat Gene 1.0 ST Array 10756687 - -
          Affymetrix Rat Gene 1.0 ST Array 10756672 - -
          Affymetrix Rat Gene 1.0 ST Array 10756680 - -
          Affymetrix Rat Gene 1.0 ST Array 10756679 - -
          Affymetrix Rat Gene 1.0 ST Array 10756686 - -
          Affymetrix Rat Gene 1.0 ST Array 10756673 - -
          Affymetrix Rat Gene 1.1 ST Array 10756671 - -
          Rat Genome 230 2.0 Array 1367551_PM_a_at - -
          Rat Genome 230 2.0 Array 1367551_a_at - -
          Rat Genome 230 2.0 Array 1392029_at - -
          Rat Genome 230 2.0 Array 1392029_PM_at - -
          Affymetrix HT Rat Focus Array 1367551_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367551_a_at - -
          Affymetrix HT Rat Focus Array 1392029_at - -
          Affymetrix HT Rat Focus Array 1392029_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001518.rn.1 TC1200000215.rn.1 RTA TC
          Rat Expression Set 230 1367551_PM_a_at - -
          Rat Expression Set 230 1367551_a_at - -
          Rat Expression Set 230 1367551_a_at - -
          Rat Expression Set 230 1392029_at - -
          Rat Expression Set 230 1367551_PM_a_at - -
          Rat Expression Set 230 1392029_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756671 - -
          Rat Genome U34 Set rc_AI169174_at - -
          Rat Genome U34 Set rc_AI175547_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21407981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353153 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-snt5b:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline