Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457371_g1
          See other UBE2A GE Assays ›
          Gene Symbol:
          Ube2a
          Gene Name:
          ubiquitin-conjugating enzyme E2A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 123486994 - 123497726 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457371_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013933.1 NP_001013955.1 1 - 2 180 97
          XM_006257446.2 XP_006257508.1 1 - 2 223 97
          GenBank mRNA BC079353.1 - 1 - 2 180 7108236 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001061.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ube2a

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          in utero embryonic development
          DNA repair
          biological_process
          positive regulation of cell proliferation
          response to UV
          protein ubiquitination
          histone ubiquitination
          histone H2A ubiquitination
          proteasome-mediated ubiquitin-dependent protein catabolic process
          protein autoubiquitination
          maternal process involved in female pregnancy
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          molecular_function
          ubiquitin-protein transferase activity
          ligase activity
          ubiquitin-like protein transferase activity
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity
          ubiquitin conjugating enzyme activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P815827 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P815827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P815827 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065957 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P815827 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P815827 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936304 - -
          Affymetrix Rat Gene 1.0 ST Array 10936305 - -
          Affymetrix Rat Gene 1.0 ST Array 10936307 - -
          Affymetrix Rat Gene 1.0 ST Array 10936310 - -
          Affymetrix Rat Gene 1.0 ST Array 10936311 - -
          Affymetrix Rat Gene 1.0 ST Array 10936309 - -
          Affymetrix Rat Gene 1.0 ST Array 10936308 - -
          Affymetrix Rat Gene 1.0 ST Array 10936306 - -
          Affymetrix Rat Gene 1.0 ST Array 10936303 - -
          Affymetrix Rat Gene 1.1 ST Array 10936302 - -
          Rat Genome 230 2.0 Array 1392573_at - -
          Rat Genome 230 2.0 Array 1392573_PM_at - -
          Rat Genome 230 2.0 Array 1388863_PM_at - -
          Rat Genome 230 2.0 Array 1388863_at - -
          Affymetrix HT Rat Focus Array 1392573_at - -
          Affymetrix HT Rat Focus Array 1388863_at - -
          Affymetrix HT Rat Focus Array 1392573_PM_at - -
          Affymetrix HT Rat Focus Array 1388863_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00006574.rn.1 TC0X00001061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00006573.rn.1 TC0X00001061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00006575.rn.1 TC0X00001061.rn.1 RTA TC
          Rat Expression Set 230 1388863_PM_at - -
          Rat Expression Set 230 1388863_at - -
          Rat Expression Set 230 1392573_at - -
          Rat Expression Set 230 1392573_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936302 - -
          Rat Genome U34 Set rc_AA956114_at - -
          Rat Genome U34 Set rc_AA956114_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22013968 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358091 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline