Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457502_m1
          See other SIRT2 GE Assays ›
          Gene Symbol:
          Sirt2
          Gene Name:
          sirtuin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 86948881 - 86971954 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457502_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sirt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008368.1 NP_001008369.1 12 - 13 1232 65
          XM_006228672.2 XP_006228734.1 11 - 12 1180 65
          XM_006228673.2 XP_006228735.1 12 - 13 1185 65
          XM_006228675.1 XP_006228737.1 12 - 13 964 65
          XM_006228676.2 XP_006228738.1 13 - 14 969 65
          GenBank mRNA BC086545.1 - 12 - 13 1232 7134377 65
          FQ212805.1 - 12 - 13 891 65
          FQ216424.1 - 8 - 9 560 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001188.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sirt2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          protein deacetylation
          autophagy
          mitotic nuclear division
          central nervous system development
          negative regulation of cell proliferation
          negative regulation of autophagy
          negative regulation of peptidyl-threonine phosphorylation
          phosphatidylinositol 3-kinase signaling
          histone deacetylation
          substantia nigra development
          myelination in peripheral nervous system
          regulation of myelination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          cellular response to oxidative stress
          peptidyl-lysine deacetylation
          cellular response to hepatocyte growth factor stimulus
          negative regulation of protein catabolic process
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA binding
          protein kinase B signaling
          cellular lipid catabolic process
          regulation of fat cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of meiotic nuclear division
          negative regulation of striated muscle tissue development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          hepatocyte growth factor receptor signaling pathway
          negative regulation of oligodendrocyte differentiation
          cell division
          meiotic cell cycle
          regulation of cell cycle
          positive regulation of cell division
          positive regulation of attachment of spindle microtubules to kinetochore
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          cellular response to caloric restriction
          negative regulation of oligodendrocyte progenitor proliferation
          histone H3 deacetylation
          histone H4 deacetylation
          cellular response to molecule of bacterial origin
          cellular response to hypoxia
          cellular response to epinephrine stimulus
          tubulin deacetylation
          positive regulation of execution phase of apoptosis
          positive regulation of oocyte maturation
          negative regulation of NLRP3 inflammasome complex assembly
          negative regulation of defense response to bacterium
          ripoptosome assembly involved in necroptotic process
          negative regulation of reactive oxygen species metabolic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia
          molecular_function
          chromatin binding
          histone deacetylase activity
          transcription factor binding
          zinc ion binding
          NAD-dependent histone deacetylase activity
          protein deacetylase activity
          NAD-dependent protein deacetylase activity
          histone acetyltransferase binding
          histone deacetylase binding
          tubulin deacetylase activity
          ubiquitin binding
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          beta-tubulin binding
          NAD+ binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0100008929.rn.1 TC0100001188.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline