Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457715_g1
          See other LOC688924 GE Assays ›
          Gene Symbol:
          LOC688924
          Gene Name:
          hypothetical protein LOC688924
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 89133158 - 89151208 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457715_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC688924
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109519.1 NP_001102989.1 3 - 4 288 82
          XM_008759189.1 XP_008757411.1 3 - 4 330 82
          XM_008759190.1 XP_008757412.1 3 - 4 318 82
          XM_008759191.1 XP_008757413.1 4 - 5 358 82
          XM_008759192.1 XP_008757414.1 4 - 5 474 82
          XM_008759193.1 XP_008757415.1 3 - 4 301 82
          XM_008759194.1 XP_008757416.1 3 - 4 393 82
          XM_008759195.1 XP_008757417.1 3 - 4 277 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001256.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P024635 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119390 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P161705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024635 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119390 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161705 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024635 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119390 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161705 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119390 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P024635 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706035 - -
          Affymetrix Rat Gene 1.0 ST Array 10706034 - -
          Affymetrix Rat Gene 1.1 ST Array 10706033 - -
          Affymetrix Rat Clariom S Assay PSR0100009782.rn.1 TC0100001256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009784.rn.1 TC0100001256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009787.rn.1 TC0100001256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009786.rn.1 TC0100001256.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10706033 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21767013 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline