Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457724_m1
          See other BCKDHA GE Assays ›
          Gene Symbol:
          Bckdha
          Gene Name:
          branched chain ketoacid dehydrogenase E1, alpha polypeptide
          Gene Aliases:
          BCKDA, E1a
          Chromosome Location:
          Chr.1: 82423291 - 82452057 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457724_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bckdha
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012782.1 NP_036914.1 5 - 6 653 94
          GenBank mRNA BC089915.1 - 3 - 4 300 94
          BC161819.1 - 5 - 6 658 9099948 94
          J02827.1 - 5 - 6 637 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005001.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bckdha

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to nutrient
          branched-chain amino acid catabolic process
          response to glucocorticoid
          response to cAMP
          oxidation-reduction process
          alpha-ketoacid dehydrogenase activity
          3-methyl-2-oxobutanoate dehydrogenase (2-methylpropanoyl-transferring) activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
          protein complex binding
          metal ion binding
          2-oxoisovalerate dehydrogenase (acylating) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832236 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59864 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15665 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15665 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15665 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700251752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64151 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94975 - -
          Agilent Rat cDNA Microarray (G4105A) 700251752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719870 - -
          Affymetrix Rat Gene 1.0 ST Array 10719869 - -
          Affymetrix Rat Gene 1.0 ST Array 10719865 - -
          Affymetrix Rat Gene 1.0 ST Array 10719866 - -
          Affymetrix Rat Gene 1.0 ST Array 10719864 - -
          Affymetrix Rat Gene 1.0 ST Array 10719871 - -
          Affymetrix Rat Gene 1.0 ST Array 10719872 - -
          Affymetrix Rat Gene 1.0 ST Array 10719867 - -
          Affymetrix Rat Gene 1.0 ST Array 10719868 - -
          Affymetrix Rat Gene 1.1 ST Array 10719863 - -
          Rat Genome 230 2.0 Array 1380557_PM_at - -
          Rat Genome 230 2.0 Array 1370897_at - -
          Rat Genome 230 2.0 Array 1370897_PM_at - -
          Rat Genome 230 2.0 Array 1382828_at - -
          Rat Genome 230 2.0 Array 1380557_at - -
          Rat Genome 230 2.0 Array 1382828_PM_at - -
          Affymetrix HT Rat Focus Array 1370897_PM_at - -
          Affymetrix HT Rat Focus Array 1380557_at - -
          Affymetrix HT Rat Focus Array 1370897_at - -
          Affymetrix HT Rat Focus Array 1380557_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100039707.rn.1 TC0100005001.rn.1 RTA TC
          Rat Expression Set 230 1370897_PM_at - -
          Rat Expression Set 230 1370897_at - -
          Rat Expression Set 230 1380557_PM_at - -
          Rat Expression Set 230 1382828_PM_at - -
          Rat Expression Set 230 1380557_at - -
          Rat Expression Set 230 1382828_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719863 - -
          Rat Genome U34 Set J02827_at - -
          Rat Genome U34 Set J02827_g_at - -
          Rat Genome U34 Set rc_AA851950_at - -
          Rat Genome U34 Set rc_AI145529_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21558354 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371691 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline