Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457880_m1
          See other CYP2B1 GE Assays ›
          Gene Symbol:
          Cyp2b1
          Gene Name:
          cytochrome P450, family 2, subfamily b, polypeptide 1
          Gene Aliases:
          Cyp2b-1, Cypbe
          Chromosome Location:
          Chr.7: 99142431 - 99183540 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457880_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134844.1 NP_001128316.1 1312 92
          XM_008765491.1 XP_008763713.1 1355 92
          XM_008765492.1 XP_008763714.1 1331 92
          XM_008765493.1 XP_008763715.1 1717 92
          GenBank mRNA BC166502.1 - 1312 7122207 92
          J00719.1 - 1280 92
          M37134.1 - 1280 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001172.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          response to metal ion
          response to sulfur dioxide
          response to organic cyclic compound
          drug metabolic process
          epoxygenase P450 pathway
          response to insulin
          response to drug
          exogenous drug catabolic process
          response to calcium ion
          oxidation-reduction process
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124515 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705228 - -
          Affymetrix Rat Gene 1.0 ST Array 10705226 - -
          Affymetrix Rat Gene 1.0 ST Array 10705229 - -
          Affymetrix Rat Gene 1.0 ST Array 10705227 - -
          Affymetrix Rat Gene 1.1 ST Array 10705225 - -
          Rat Genome 230 2.0 Array 1371076_PM_at - -
          Rat Genome 230 2.0 Array 1371076_at - -
          Affymetrix Rat Clariom S Assay PSR0700008592.rn.1 TC0700001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700008593.rn.1 TC0700001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700008594.rn.1 TC0700001172.rn.1 RTA TC
          Rat Expression Set 230 1371076_PM_at - -
          Rat Expression Set 230 1371076_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705225 - -
          Rat Genome U34 Set M11251cds_f_at - -
          Rat Genome U34 Set K00996mRNA_s_at - -
          Rat Genome U34 Set L00320cds_f_at - -
          Rat Genome U34 Set rc_AA818412_f_at - -
          Rat Genome U34 Set L00320cds_f_at - -
          Rat Genome U34 Set M11251cds_f_at - -
          Rat Genome U34 Set K00996mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21240354 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline