Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458013_m1
          See other ZFP180 GE Assays ›
          Gene Symbol:
          Zfp180
          Gene Name:
          zinc finger protein 180
          Gene Aliases:
          rKr1
          Chromosome Location:
          Chr.1: 80920745 - 80940038 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01458013_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp180
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271280.1 NP_001258209.1 4 - 5 625 73
          NM_144757.3 NP_653358.2 4 - 5 543 73
          XM_006228386.2 XP_006228448.1 4 - 5 576 73
          XM_006228387.2 XP_006228449.1 4 - 5 627 73
          GenBank mRNA U41164.1 - 4 - 5 543 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001024.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp180

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P332310 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P332310 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P332310 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101339 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704896 - -
          Affymetrix Rat Gene 1.0 ST Array 10704900 - -
          Affymetrix Rat Gene 1.0 ST Array 10704899 - -
          Affymetrix Rat Gene 1.0 ST Array 10704901 - -
          Affymetrix Rat Gene 1.0 ST Array 10704898 - -
          Affymetrix Rat Gene 1.1 ST Array 10704895 - -
          Rat Genome 230 2.0 Array 1387962_at - -
          Rat Genome 230 2.0 Array 1387962_PM_at - -
          Rat Genome 230 2.0 Array 1388779_PM_at - -
          Rat Genome 230 2.0 Array 1388779_at - -
          Affymetrix HT Rat Focus Array 1388779_PM_at - -
          Affymetrix HT Rat Focus Array 1387962_PM_at - -
          Affymetrix HT Rat Focus Array 1387962_at - -
          Affymetrix HT Rat Focus Array 1388779_at - -
          Affymetrix Rat Clariom S Assay PSR0100007209.rn.1 TC0100001024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007210.rn.1 TC0100001024.rn.1 RTA TC
          Rat Expression Set 230 1388779_PM_at - -
          Rat Expression Set 230 1388779_at - -
          Rat Expression Set 230 1387962_PM_at - -
          Rat Expression Set 230 1387962_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704895 - -
          Rat Genome U34 Set U41164_at - -
          Rat Genome U34 Set rc_AI058859_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21531184 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370049 - -
          RatRef-12 Expression BeadChip ILMN_58727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0