Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458152_m1
          See other KCTD15 GE Assays ›
          Gene Symbol:
          Kctd15
          Gene Name:
          potassium channel tetramerization domain containing 15
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 90506697 - 90522224 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01458152_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kctd15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109141.1 NP_001102611.1 5 - 6 978 60
          XM_006228859.2 XP_006228921.1 6 - 7 829 60
          XM_006228860.2 XP_006228922.1 6 - 7 1082 60
          XM_008759178.1 XP_008757400.1 6 - 7 904 60
          GenBank mRNA BC168920.1 - 5 - 6 922 9100405 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005207.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s)

          protein homooligomerization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628147 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142744 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54169 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721039 - -
          Affymetrix Rat Gene 1.0 ST Array 10721042 - -
          Affymetrix Rat Gene 1.0 ST Array 10721040 - -
          Affymetrix Rat Gene 1.0 ST Array 10721041 - -
          Affymetrix Rat Gene 1.0 ST Array 10721043 - -
          Affymetrix Rat Gene 1.0 ST Array 10721044 - -
          Affymetrix Rat Gene 1.1 ST Array 10721038 - -
          Rat Genome 230 2.0 Array 1372398_PM_at - -
          Rat Genome 230 2.0 Array 1372398_at - -
          Affymetrix HT Rat Focus Array 1372398_at - -
          Affymetrix HT Rat Focus Array 1372398_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100042141.rn.1 TC0100005207.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042139.rn.1 TC0100005207.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042137.rn.1 TC0100005207.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042140.rn.1 TC0100005207.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042136.rn.1 TC0100005207.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042138.rn.1 TC0100005207.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042143.rn.1 TC0100005207.rn.1 RTA TC
          Rat Expression Set 230 1372398_PM_at - -
          Rat Expression Set 230 1372398_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721038 - -
          Rat Genome U34 Set rc_H31964_at - -
          Rat Genome U34 Set rc_AI011627_at - -
          Rat Genome U34 Set rc_AA963162_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21286230 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039030 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline