Search
Search
| Gene Symbol: Dcaf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173558.1 | NP_001167029.1 | 4 - 5 | 530 | 73 | |
| XM_006241605.2 | XP_006241667.1 | 4 - 5 | 489 | 73 | ||
| GenBank mRNA | BC158730.1 | - | 2 - 3 | 124 | 73 | |
| EV763930.1 | - | 4 - 5 | 530 | 73 | ||
| FQ228874.1 | - | 4 - 5 | 540 | 73 | ||
| FQ229670.1 | - | 2 - 3 | 189 | 73 | ||
Set Membership: |
|