Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458894_m1
          See other ILF2 GE Assays ›
          Gene Symbol:
          Ilf2
          Gene Name:
          interleukin enhancer binding factor 2
          Gene Aliases:
          Ab1-143
          Chromosome Location:
          Chr.2: 189857587 - 189877777 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01458894_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ilf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047886.1 NP_001041351.1 6 - 7 525 73
          XM_006232624.2 XP_006232686.1 3 - 4 236 73
          GenBank mRNA AY325142.1 - 6 - 7 525 73
          BC167105.1 - 3 - 4 136 9102039 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001610.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          immune response
          positive regulation of transcription, DNA-templated
          DNA binding
          double-stranded RNA binding
          ATP binding
          transferase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P419107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81471 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P419107 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P419107 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P419107 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P419107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1880 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817024 - -
          Affymetrix Rat Gene 1.0 ST Array 10817030 - -
          Affymetrix Rat Gene 1.0 ST Array 10817033 - -
          Affymetrix Rat Gene 1.0 ST Array 10817026 - -
          Affymetrix Rat Gene 1.0 ST Array 10817037 - -
          Affymetrix Rat Gene 1.0 ST Array 10817028 - -
          Affymetrix Rat Gene 1.0 ST Array 10817036 - -
          Affymetrix Rat Gene 1.0 ST Array 10817032 - -
          Affymetrix Rat Gene 1.0 ST Array 10817038 - -
          Affymetrix Rat Gene 1.0 ST Array 10817025 - -
          Affymetrix Rat Gene 1.0 ST Array 10817027 - -
          Affymetrix Rat Gene 1.0 ST Array 10817035 - -
          Affymetrix Rat Gene 1.0 ST Array 10817023 - -
          Affymetrix Rat Gene 1.0 ST Array 10817034 - -
          Affymetrix Rat Gene 1.0 ST Array 10817022 - -
          Affymetrix Rat Gene 1.0 ST Array 10817039 - -
          Affymetrix Rat Gene 1.0 ST Array 10817031 - -
          Affymetrix Rat Gene 1.0 ST Array 10817029 - -
          Affymetrix Rat Gene 1.1 ST Array 10817021 - -
          Rat Genome 230 2.0 Array 1371582_at - -
          Rat Genome 230 2.0 Array 1371582_PM_at - -
          Affymetrix HT Rat Focus Array 1371582_at - -
          Affymetrix HT Rat Focus Array 1371582_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200010771.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010802.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010796.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010768.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010769.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010772.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010798.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010801.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010804.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010805.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010789.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010797.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010783.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010795.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010770.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010784.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010794.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010786.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010792.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010799.rn.1 TC0200001610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200010782.rn.1 TC0200001610.rn.1 RTA TC
          Rat Expression Set 230 1371582_at - -
          Rat Expression Set 230 1371582_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817021 - -
          Rat Genome U34 Set rc_AA848402_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21976706 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355274 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline