Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459028_m1
          See other FGB GE Assays ›
          Gene Symbol:
          Fgb
          Gene Name:
          fibrinogen beta chain
          Gene Aliases:
          Ab1-181, Ab1-216, Ac1-581
          Chromosome Location:
          Chr.2: 182027100 - 182037123 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01459028_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020071.2 NP_064456.2 3 - 4 491 154
          XM_006232524.2 XP_006232586.1 3 - 4 509 154
          XM_008761078.1 XP_008759300.1 3 - 4 448 154
          GenBank mRNA AY321323.1 - 487 154
          AY325147.1 - 487 154
          AY325153.1 - 487 154
          BC088102.1 - 3 - 4 491 7371665 154
          FQ209924.1 - 3 - 4 498 154
          FQ219024.1 - 3 - 4 401 154
          U05675.1 - 3 - 4 492 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003941.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgb

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          signal transduction
          positive regulation of heterotypic cell-cell adhesion
          cellular protein complex assembly
          cellular response to leptin stimulus
          innate immune response
          positive regulation of vasoconstriction
          positive regulation of exocytosis
          positive regulation of protein secretion
          protein polymerization
          response to calcium ion
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          cellular response to interleukin-1
          positive regulation of peptide hormone secretion
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          receptor binding
          structural molecule activity
          protein binding, bridging
          cell adhesion molecule binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P070791 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10037 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P070791 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10037 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226859 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1044835 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10037 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700606793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42767 - -
          Agilent Rat cDNA Microarray (G4105A) 700606793 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823965 - -
          Affymetrix Rat Gene 1.0 ST Array 10823963 - -
          Affymetrix Rat Gene 1.0 ST Array 10823964 - -
          Affymetrix Rat Gene 1.0 ST Array 10823961 - -
          Affymetrix Rat Gene 1.0 ST Array 10823958 - -
          Affymetrix Rat Gene 1.0 ST Array 10823957 - -
          Affymetrix Rat Gene 1.0 ST Array 10823962 - -
          Affymetrix Rat Gene 1.0 ST Array 10823960 - -
          Affymetrix Rat Gene 1.0 ST Array 10823959 - -
          Affymetrix Rat Gene 1.1 ST Array 10823955 - -
          Rat Genome 230 2.0 Array 1371234_PM_at - -
          Rat Genome 230 2.0 Array 1388266_at - -
          Rat Genome 230 2.0 Array 1371234_at - -
          Rat Genome 230 2.0 Array 1388266_PM_at - -
          Rat Genome 230 2.0 Array 1370511_at - -
          Rat Genome 230 2.0 Array 1370511_PM_at - -
          Affymetrix HT Rat Focus Array 1370511_at - -
          Affymetrix HT Rat Focus Array 1388266_PM_at - -
          Affymetrix HT Rat Focus Array 1371234_at - -
          Affymetrix HT Rat Focus Array 1388266_at - -
          Affymetrix HT Rat Focus Array 1370511_PM_at - -
          Affymetrix HT Rat Focus Array 1371234_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200026482.rn.1 TC0200003941.rn.1 RTA TC
          Rat Expression Set 230 1371234_at - -
          Rat Expression Set 230 1388266_at - -
          Rat Expression Set 230 1370511_PM_at - -
          Rat Expression Set 230 1370511_at - -
          Rat Expression Set 230 1388266_PM_at - -
          Rat Expression Set 230 1371234_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823955 - -
          Rat Genome U34 Set U05675_at - -
          Rat Genome U34 Set M35602_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21964976 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354112 - -
          RatRef-12 Expression BeadChip ILMN_47985 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline