Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459065_g1
          See other HAPLN2 GE Assays ›
          Gene Symbol:
          Hapln2
          Gene Name:
          hyaluronan and proteoglycan link protein 2
          Gene Aliases:
          Bral1
          Chromosome Location:
          Chr.2: 187395104 - 187404813 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01459065_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hapln2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022285.1 NP_071621.1 6 - 7 936 59
          XM_006232751.2 XP_006232813.1 6 - 7 1018 59
          XM_008761208.1 XP_008759430.1 7 - 8 1059 59
          XM_008761209.1 XP_008759431.1 6 - 7 1244 59
          GenBank mRNA AB049056.1 - 6 - 7 936 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004018.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hapln2

          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          cell adhesion
          central nervous system development
          establishment of blood-nerve barrier
          extracellular matrix assembly
          extracellular matrix structural constituent
          hyaluronic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110024 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141732 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141732 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15779 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P203355 - -
          Agilent Rat cDNA Microarray (G4105A) 701746930 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824246 - -
          Affymetrix Rat Gene 1.0 ST Array 10824248 - -
          Affymetrix Rat Gene 1.0 ST Array 10824247 - -
          Affymetrix Rat Gene 1.0 ST Array 10824244 - -
          Affymetrix Rat Gene 1.0 ST Array 10824249 - -
          Affymetrix Rat Gene 1.0 ST Array 10824245 - -
          Affymetrix Rat Gene 1.0 ST Array 10824243 - -
          Affymetrix Rat Gene 1.1 ST Array 10824242 - -
          Rat Genome 230 2.0 Array 1390017_at - -
          Rat Genome 230 2.0 Array 1394952_at - -
          Rat Genome 230 2.0 Array 1370849_at - -
          Rat Genome 230 2.0 Array 1390017_PM_at - -
          Rat Genome 230 2.0 Array 1394952_PM_at - -
          Rat Genome 230 2.0 Array 1370849_PM_at - -
          Affymetrix HT Rat Focus Array 1370849_PM_at - -
          Affymetrix HT Rat Focus Array 1394952_PM_at - -
          Affymetrix HT Rat Focus Array 1370849_at - -
          Affymetrix HT Rat Focus Array 1390017_at - -
          Affymetrix HT Rat Focus Array 1394952_at - -
          Affymetrix HT Rat Focus Array 1390017_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200027138.rn.1 TC0200004018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027139.rn.1 TC0200004018.rn.1 RTA TC
          Rat Expression Set 230 1390017_at - -
          Rat Expression Set 230 1390017_PM_at - -
          Rat Expression Set 230 1370849_PM_at - -
          Rat Expression Set 230 1370849_at - -
          Rat Expression Set 230 1394952_at - -
          Rat Expression Set 230 1394952_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824242 - -
          Rat Genome U34 Set rc_AI145465_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21734028 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357365 - -
          RatRef-12 Expression BeadChip ILMN_69380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline