Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459299_m1
          See other FKBP4 GE Assays ›
          Gene Symbol:
          Fkbp4
          Gene Name:
          FK506 binding protein 4
          Gene Aliases:
          FKBP-4, FKBP-52, p59
          Chromosome Location:
          Chr.4: 161748993 - 161757447 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01459299_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191863.1 NP_001178792.1 1 - 2 265 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003829.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          steroid hormone receptor complex assembly
          copper ion transport
          embryo implantation
          negative regulation of neuron projection development
          androgen receptor signaling pathway
          prostate gland development
          negative regulation of microtubule polymerization or depolymerization
          negative regulation of microtubule polymerization
          protein complex localization
          male sex differentiation
          reproductive structure development
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          ATP binding
          GTP binding
          FK506 binding
          heat shock protein binding
          copper-dependent protein binding
          glucocorticoid receptor binding
          poly(A) RNA binding
          tau protein binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P179282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P179282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P179282 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P179282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139925 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P439665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865862 - -
          Affymetrix Rat Gene 1.0 ST Array 10865860 - -
          Affymetrix Rat Gene 1.0 ST Array 10865864 - -
          Affymetrix Rat Gene 1.0 ST Array 10865858 - -
          Affymetrix Rat Gene 1.0 ST Array 10865857 - -
          Affymetrix Rat Gene 1.0 ST Array 10865856 - -
          Affymetrix Rat Gene 1.0 ST Array 10865859 - -
          Affymetrix Rat Gene 1.0 ST Array 10865861 - -
          Affymetrix Rat Gene 1.0 ST Array 10865863 - -
          Affymetrix Rat Gene 1.1 ST Array 10865855 - -
          Rat Genome 230 2.0 Array 1389844_at - -
          Rat Genome 230 2.0 Array 1389844_PM_at - -
          Affymetrix HT Rat Focus Array 1389844_at - -
          Affymetrix HT Rat Focus Array 1389844_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400027191.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027185.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027188.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027190.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027195.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027193.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027184.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027187.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027192.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027196.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027194.rn.1 TC0400003829.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027186.rn.1 TC0400003829.rn.1 RTA TC
          Rat Expression Set 230 1389844_PM_at - -
          Rat Expression Set 230 1389844_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865855 - -
          Rat Genome U34 Set rc_AI136977_at - -
          Rat Genome U34 Set rc_AI231547_at - -
          Rat Genome U34 Set rc_AI136977_g_at - -
          Rat Genome U34 Set rc_AI012177_at - -
          Rat Genome U34 Set rc_AI231547_at - -
          Rat Genome U34 Set rc_AI136977_g_at - -
          Rat Genome U34 Set rc_AI136977_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358928 - -
          RatRef-12 Expression BeadChip ILMN_57582 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline