Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459418_m1
          See other CLEC4D GE Assays ›
          Gene Symbol:
          Clec4d
          Gene Name:
          C-type lectin domain family 4, member D
          Gene Aliases:
          Clecsf8, Mcl
          Chromosome Location:
          Chr.4: 156253084 - 156262644 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01459418_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec4d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001003707.1 NP_001003707.1 5 - 6 531 88
          XM_006237322.2 XP_006237384.1 5 - 6 713 88
          GenBank mRNA AY339882.1 - 5 - 6 531 88
          AY494065.1 - 5 - 6 531 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001738.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec4d

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P388783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P388783 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P388783 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P388783 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104968 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858600 - -
          Affymetrix Rat Gene 1.0 ST Array 10858605 - -
          Affymetrix Rat Gene 1.0 ST Array 10858603 - -
          Affymetrix Rat Gene 1.0 ST Array 10858604 - -
          Affymetrix Rat Gene 1.0 ST Array 10858601 - -
          Affymetrix Rat Gene 1.0 ST Array 10858602 - -
          Affymetrix Rat Gene 1.1 ST Array 10858599 - -
          Affymetrix Rat Clariom S Assay PSR0400012636.rn.1 TC0400001738.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012635.rn.1 TC0400001738.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10858599 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21085505 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline