Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459492_m1
          See other ERGIC2 GE Assays ›
          Gene Symbol:
          Ergic2
          Gene Name:
          ERGIC and golgi 2
          Gene Aliases:
          RGD1310606
          Chromosome Location:
          Chr.4: 182569086 - 182600258 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01459492_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ergic2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024984.1 NP_001020155.1 9 - 10 837 134
          GenBank mRNA BC083597.1 - 9 - 10 837 7132841 134
          FQ213743.1 - 10 - 11 804 134
          FQ227558.1 - 9 - 10 722 134
          FQ230170.1 - 11 - 12 920 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004116.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ergic2

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P420646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106373 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P420646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P420646 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P420646 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P420646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51164 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3700 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866983 - -
          Affymetrix Rat Gene 1.0 ST Array 10866978 - -
          Affymetrix Rat Gene 1.0 ST Array 10866980 - -
          Affymetrix Rat Gene 1.0 ST Array 10866975 - -
          Affymetrix Rat Gene 1.0 ST Array 10866982 - -
          Affymetrix Rat Gene 1.0 ST Array 10866974 - -
          Affymetrix Rat Gene 1.0 ST Array 10866981 - -
          Affymetrix Rat Gene 1.0 ST Array 10866977 - -
          Affymetrix Rat Gene 1.0 ST Array 10866973 - -
          Affymetrix Rat Gene 1.0 ST Array 10866984 - -
          Affymetrix Rat Gene 1.0 ST Array 10866985 - -
          Affymetrix Rat Gene 1.1 ST Array 10866972 - -
          Rat Genome 230 2.0 Array 1389970_at - -
          Rat Genome 230 2.0 Array 1386266_at - -
          Rat Genome 230 2.0 Array 1389970_PM_at - -
          Rat Genome 230 2.0 Array 1386266_PM_at - -
          Affymetrix HT Rat Focus Array 1389970_PM_at - -
          Affymetrix HT Rat Focus Array 1389970_at - -
          Affymetrix Rat Clariom S Assay PSR0400029230.rn.1 TC0400004116.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029231.rn.1 TC0400004116.rn.1 RTA TC
          Rat Expression Set 230 1389970_PM_at - -
          Rat Expression Set 230 1389970_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866972 - -
          Rat Genome U34 Set rc_AA964275_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21660821 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354609 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline